Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61118150-61118395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD5
Entrez Gene ID
921
Gene Name
CD5 molecule
Gene Aliases
LEU1, T1
UniGene
Hs.58685
Gene Chromosome Location
Chr.11: 61102458-61127852 on Build GRCh38
Transcripts
NM_014207.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6891175 c.150C>T p.V50V Chr.11: 61118230-61118230 on Build GRCh38
5798263 c.207G>C p.K69N Chr.11: 61118287-61118287 on Build GRCh38
689349 c.108G>A p.R36R Chr.11: 61118188-61118188 on Build GRCh38
5959220 c.202T>G p.S68A Chr.11: 61118282-61118282 on Build GRCh38
4034550 c.232T>C p.S78P Chr.11: 61118312-61118312 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766117426 Chr.11: 61118255-61118255 on Build GRCh38
rs748162258 Chr.11: 61118331-61118331 on Build GRCh38
rs773124953 Chr.11: 61118356-61118356 on Build GRCh38
rs769791636 Chr.11: 61118340-61118340 on Build GRCh38
rs149268422 Chr.11: 61118277-61118277 on Build GRCh38
rs182453573 Chr.11: 61118253-61118253 on Build GRCh38
rs751153028 Chr.11: 61118180-61118180 on Build GRCh38
rs747163529 Chr.11: 61118323-61118323 on Build GRCh38
rs757852160 Chr.11: 61118283-61118283 on Build GRCh38
rs959799700 Chr.11: 61118215-61118215 on Build GRCh38
rs373005084 Chr.11: 61118236-61118236 on Build GRCh38
rs1002727536 Chr.11: 61118361-61118361 on Build GRCh38
rs918103823 Chr.11: 61118235-61118235 on Build GRCh38
rs562627397 Chr.11: 61118282-61118282 on Build GRCh38
rs34315555 Chr.11: 61118181-61118182 on Build GRCh38
rs370429428 Chr.11: 61118272-61118272 on Build GRCh38
rs749160317 Chr.11: 61118372-61118372 on Build GRCh38
rs138549146 Chr.11: 61118206-61118206 on Build GRCh38
rs746175908 Chr.11: 61118205-61118205 on Build GRCh38
rs758676539 Chr.11: 61118299-61118299 on Build GRCh38
rs144361212 Chr.11: 61118311-61118311 on Build GRCh38
rs138915999 Chr.11: 61118328-61118328 on Build GRCh38
rs922916722 Chr.11: 61118194-61118194 on Build GRCh38
rs146299312 Chr.11: 61118250-61118250 on Build GRCh38
rs141935047 Chr.11: 61118188-61118188 on Build GRCh38
rs750884036 Chr.11: 61118296-61118296 on Build GRCh38
rs747170921 Chr.11: 61118214-61118214 on Build GRCh38
rs780135342 Chr.11: 61118209-61118209 on Build GRCh38
rs755184614 Chr.11: 61118327-61118327 on Build GRCh38
rs887498822 Chr.11: 61118274-61118274 on Build GRCh38
rs768592268 Chr.11: 61118222-61118222 on Build GRCh38
rs369731937 Chr.11: 61118203-61118203 on Build GRCh38
rs777739167 Chr.11: 61118202-61118204 on Build GRCh38
rs200568806 Chr.11: 61118171-61118171 on Build GRCh38
rs369225298 Chr.11: 61118232-61118232 on Build GRCh38
rs765770381 Chr.11: 61118291-61118291 on Build GRCh38
rs772998606 Chr.11: 61118249-61118249 on Build GRCh38
rs766968428 Chr.11: 61118280-61118280 on Build GRCh38
rs765995942 Chr.11: 61118307-61118307 on Build GRCh38
rs765883722 Chr.11: 61118173-61118173 on Build GRCh38
rs754367132 Chr.11: 61118193-61118193 on Build GRCh38
rs150082740 Chr.11: 61118196-61118196 on Build GRCh38
rs769667296 Chr.11: 61118243-61118243 on Build GRCh38
rs759127306 Chr.11: 61118276-61118276 on Build GRCh38
rs1027275648 Chr.11: 61118181-61118181 on Build GRCh38
rs761767751 Chr.11: 61118242-61118242 on Build GRCh38

View Full Product Details