Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14670909-14671166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC174
Entrez Gene ID
51244
Gene Name
coiled-coil domain containing 174
Gene Aliases
C3orf19, HSPC212, IHPM, IHPMR
UniGene
Hs.517820
Gene Chromosome Location
Chr.3: 14651683-14673186 on Build GRCh38
Transcripts
NM_016474.4 NR_135523.1 XM_017006554.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3002210 c.1359C>A p.F453L Chr.3: 14671149-14671149 on Build GRCh38
1471564 c.1375A>G p.M459V Chr.3: 14671165-14671165 on Build GRCh38
1180160 c.1218C>T p.S406S Chr.3: 14671008-14671008 on Build GRCh38
390607 c.1127C>A p.S376* Chr.3: 14670917-14670917 on Build GRCh38
7314090 c.1195G>T p.G399C Chr.3: 14670985-14670985 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751625276 Chr.3: 14670937-14670937 on Build GRCh38
rs753254506 Chr.3: 14671008-14671008 on Build GRCh38
rs145581381 Chr.3: 14670941-14670941 on Build GRCh38
rs755519616 Chr.3: 14670938-14670938 on Build GRCh38
rs942928321 Chr.3: 14671035-14671035 on Build GRCh38
rs759690754 Chr.3: 14671094-14671094 on Build GRCh38
rs375688410 Chr.3: 14670965-14670965 on Build GRCh38
rs1053419364 Chr.3: 14671136-14671136 on Build GRCh38
rs766113960 Chr.3: 14671092-14671092 on Build GRCh38
rs138693361 Chr.3: 14671108-14671108 on Build GRCh38
rs766459634 Chr.3: 14670935-14670935 on Build GRCh38
rs768310966 Chr.3: 14671058-14671058 on Build GRCh38
rs138553563 Chr.3: 14671080-14671080 on Build GRCh38
rs776211840 Chr.3: 14671074-14671074 on Build GRCh38
rs748606046 Chr.3: 14670957-14670957 on Build GRCh38
rs773126505 Chr.3: 14671078-14671078 on Build GRCh38
rs779827041 Chr.3: 14671057-14671057 on Build GRCh38
rs758160957 Chr.3: 14671056-14671056 on Build GRCh38
rs752710100 Chr.3: 14671098-14671098 on Build GRCh38
rs372533633 Chr.3: 14670990-14670990 on Build GRCh38
rs774588314 Chr.3: 14671093-14671093 on Build GRCh38
rs757715514 Chr.3: 14671112-14671112 on Build GRCh38
rs769604455 Chr.3: 14671077-14671077 on Build GRCh38
rs778261389 Chr.3: 14671017-14671017 on Build GRCh38
rs775202853 Chr.3: 14670968-14670968 on Build GRCh38
rs140448070 Chr.3: 14671075-14671075 on Build GRCh38
rs145950373 Chr.3: 14671103-14671103 on Build GRCh38
rs374798787 Chr.3: 14670969-14670969 on Build GRCh38
rs911230768 Chr.3: 14671020-14671020 on Build GRCh38
rs776729366 Chr.3: 14670972-14670972 on Build GRCh38
rs376403526 Chr.3: 14670986-14670986 on Build GRCh38
rs754618667 Chr.3: 14671120-14671120 on Build GRCh38
rs756523348 Chr.3: 14671010-14671010 on Build GRCh38
rs768760183 Chr.3: 14670971-14670971 on Build GRCh38
rs756535177 Chr.3: 14670964-14670964 on Build GRCh38
rs558741704 Chr.3: 14671051-14671051 on Build GRCh38
rs367970098 Chr.3: 14670973-14670973 on Build GRCh38
rs368635335 Chr.3: 14671133-14671133 on Build GRCh38
rs772824290 Chr.3: 14670980-14670980 on Build GRCh38
rs755074520 Chr.3: 14671001-14671001 on Build GRCh38
rs750686787 Chr.3: 14670932-14670932 on Build GRCh38
rs1901 Chr.3: 14670966-14670966 on Build GRCh38
rs538749910 Chr.3: 14670982-14670982 on Build GRCh38
rs573969015 Chr.3: 14671117-14671117 on Build GRCh38
rs374228927 Chr.3: 14671116-14671116 on Build GRCh38
rs148416293 Chr.3: 14671100-14671100 on Build GRCh38
rs60239620 Chr.3: 14671097-14671097 on Build GRCh38

View Full Product Details