Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 40821840-40822091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBB2
Entrez Gene ID
323
Gene Name
amyloid beta precursor protein binding family B member 2
Gene Aliases
FE65L, FE65L1
UniGene
Hs.479602
Gene Chromosome Location
Chr.4: 40810027-41216714 on Build GRCh38
Transcripts
NM_001166050.1 NM_001166051.1 NM_001166052.1 NM_001166053.1 NM_001166054.1 NM_004307.1 NM_173075.4 XM_005248101.3 XM_006714005.3 XM_006714007.3 XM_006714008.3 XM_006714009.3 XM_006714010.3 XM_006714011.3 XM_006714012.3 XM_011513686.2 XM_011513687.2 XM_017008142.1 XM_017008143.1 XM_017008144.1 XM_017008145.1 XM_017008146.1 XM_017008147.1 XM_017008148.1 XM_017008149.1 XM_017008150.1 XM_017008151.1 XM_017008152.1 XM_017008153.1 XM_017008154.1 XM_017008155.1 XM_017008156.1 XM_017008157.1 XM_017008158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5355846 c.2036G>A p.R679H Chr.4: 40821944-40821944 on Build GRCh38
367348 c.1951A>G p.M651V Chr.4: 40821963-40821963 on Build GRCh38
4423884 c.1964G>A p.R655Q Chr.4: 40822016-40822016 on Build GRCh38
4124686 c.1929C>T p.D643D Chr.4: 40821985-40821985 on Build GRCh38
5677871 c.2056T>G p.W686G Chr.4: 40821924-40821924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964980026 Chr.4: 40821886-40821886 on Build GRCh38
rs753735850 Chr.4: 40822010-40822010 on Build GRCh38
rs768813900 Chr.4: 40821958-40821958 on Build GRCh38
rs780699277 Chr.4: 40822043-40822049 on Build GRCh38
rs371307741 Chr.4: 40821944-40821944 on Build GRCh38
rs764875581 Chr.4: 40822045-40822045 on Build GRCh38
rs953905646 Chr.4: 40821984-40821984 on Build GRCh38
rs758056722 Chr.4: 40822036-40822036 on Build GRCh38
rs758087545 Chr.4: 40821933-40821933 on Build GRCh38
rs770069501 Chr.4: 40821859-40821859 on Build GRCh38
rs763192901 Chr.4: 40821956-40821956 on Build GRCh38
rs775674243 Chr.4: 40821880-40821880 on Build GRCh38
rs763101091 Chr.4: 40821882-40821882 on Build GRCh38
rs778306463 Chr.4: 40821934-40821934 on Build GRCh38
rs777536990 Chr.4: 40822066-40822066 on Build GRCh38
rs752208971 Chr.4: 40822048-40822048 on Build GRCh38
rs113282369 Chr.4: 40821889-40821889 on Build GRCh38
rs747314373 Chr.4: 40821937-40821937 on Build GRCh38
rs895204989 Chr.4: 40821861-40821861 on Build GRCh38
rs532403328 Chr.4: 40822037-40822037 on Build GRCh38
rs763868115 Chr.4: 40822067-40822067 on Build GRCh38
rs147543620 Chr.4: 40822068-40822068 on Build GRCh38
rs376616524 Chr.4: 40821919-40821919 on Build GRCh38
rs750178135 Chr.4: 40821900-40821900 on Build GRCh38
rs781561102 Chr.4: 40822069-40822069 on Build GRCh38
rs759482886 Chr.4: 40822039-40822039 on Build GRCh38
rs976977105 Chr.4: 40821972-40821972 on Build GRCh38
rs377623770 Chr.4: 40821943-40821943 on Build GRCh38
rs114406176 Chr.4: 40821945-40821945 on Build GRCh38
rs760354590 Chr.4: 40821985-40821985 on Build GRCh38
rs191884680 Chr.4: 40821930-40821930 on Build GRCh38
rs769873401 Chr.4: 40821955-40821955 on Build GRCh38
rs758173521 Chr.4: 40822062-40822062 on Build GRCh38
rs766403895 Chr.4: 40822002-40822002 on Build GRCh38
rs369075061 Chr.4: 40821858-40821858 on Build GRCh38
rs374152648 Chr.4: 40821931-40821931 on Build GRCh38
rs754438516 Chr.4: 40821922-40821922 on Build GRCh38
rs548111352 Chr.4: 40821901-40821901 on Build GRCh38
rs773104241 Chr.4: 40821977-40821977 on Build GRCh38
rs963099402 Chr.4: 40821967-40821967 on Build GRCh38
rs746017046 Chr.4: 40821949-40821949 on Build GRCh38

View Full Product Details