Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232768057-232768310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIGYF2
Entrez Gene ID
26058
Gene Name
GRB10 interacting GYF protein 2
Gene Aliases
GYF2, PARK11, PERQ2, PERQ3, TNRC15
UniGene
Hs.565319
Gene Chromosome Location
Chr.2: 232697305-232860577 on Build GRCh38
Transcripts
NM_001103146.1 NM_001103147.1 NM_001103148.1 NM_015575.3 NR_103492.1 NM_001172416.1 NM_001172417.1 NM_002242.4

Gene Symbol
KCNJ13
Entrez Gene ID
3769
Gene Name
potassium voltage-gated channel subfamily J member 13
Gene Aliases
KIR1.4, KIR7.1, LCA16, SVD
UniGene
Hs.467338
Gene Chromosome Location
Chr.2: 232765802-232776565 on Build GRCh38
Transcripts
NM_001103146.1 NM_001103147.1 NM_001103148.1 NM_015575.3 NR_103492.1 NM_001172416.1 NM_001172417.1 NM_002242.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7194901 c.1030A>T p.N344Y Chr.2: 232768244-232768244 on Build GRCh38
4933062 c.986T>C p.L329P Chr.2: 232768288-232768288 on Build GRCh38
1662366 c.1050T>A p.N350K Chr.2: 232768224-232768224 on Build GRCh38
3407670 c.1032T>C p.N344N Chr.2: 232768242-232768242 on Build GRCh38
3838852 c.1059C>A p.I353I Chr.2: 232768215-232768215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759022075 Chr.2: 232768161-232768161 on Build GRCh38
rs757634383 Chr.2: 232768274-232768274 on Build GRCh38
rs570138880 Chr.2: 232768097-232768097 on Build GRCh38
rs769427290 Chr.2: 232768189-232768189 on Build GRCh38
rs752863162 Chr.2: 232768260-232768260 on Build GRCh38
rs767179140 Chr.2: 232768144-232768144 on Build GRCh38
rs886055795 Chr.2: 232768089-232768089 on Build GRCh38
rs755099409 Chr.2: 232768154-232768154 on Build GRCh38
rs777663064 Chr.2: 232768267-232768267 on Build GRCh38
rs765534142 Chr.2: 232768253-232768253 on Build GRCh38
rs374301010 Chr.2: 232768184-232768184 on Build GRCh38
rs779138733 Chr.2: 232768157-232768157 on Build GRCh38
rs778392451 Chr.2: 232768168-232768168 on Build GRCh38
rs535054686 Chr.2: 232768198-232768198 on Build GRCh38
rs1004804255 Chr.2: 232768173-232768173 on Build GRCh38
rs570363677 Chr.2: 232768242-232768242 on Build GRCh38
rs750120231 Chr.2: 232768233-232768233 on Build GRCh38
rs888593629 Chr.2: 232768156-232768156 on Build GRCh38
rs376837580 Chr.2: 232768153-232768153 on Build GRCh38
rs748224832 Chr.2: 232768159-232768159 on Build GRCh38
rs904905317 Chr.2: 232768081-232768081 on Build GRCh38
rs774380473 Chr.2: 232768218-232768218 on Build GRCh38
rs757048919 Chr.2: 232768142-232768142 on Build GRCh38
rs369373214 Chr.2: 232768143-232768143 on Build GRCh38
rs764061215 Chr.2: 232768210-232768210 on Build GRCh38
rs781665941 Chr.2: 232768276-232768276 on Build GRCh38
rs576752433 Chr.2: 232768195-232768195 on Build GRCh38
rs758581367 Chr.2: 232768263-232768263 on Build GRCh38
rs373596360 Chr.2: 232768146-232768146 on Build GRCh38
rs937752468 Chr.2: 232768089-232768089 on Build GRCh38
rs1048157859 Chr.2: 232768123-232768123 on Build GRCh38
rs747432883 Chr.2: 232768171-232768171 on Build GRCh38
rs566257347 Chr.2: 232768180-232768180 on Build GRCh38
rs771346059 Chr.2: 232768174-232768174 on Build GRCh38
rs747562741 Chr.2: 232768269-232768269 on Build GRCh38
rs545594184 Chr.2: 232768231-232768231 on Build GRCh38
rs942173837 Chr.2: 232768105-232768105 on Build GRCh38
rs370090481 Chr.2: 232768177-232768177 on Build GRCh38
rs762432264 Chr.2: 232768209-232768209 on Build GRCh38

View Full Product Details