Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18015993-18016266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATPAF2
Entrez Gene ID
91647
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 2
Gene Aliases
ATP12, ATP12p, LP3663, MC5DN1
UniGene
Hs.528889
Gene Chromosome Location
Chr.17: 18015059-18039169 on Build GRCh38
Transcripts
XM_011524065.1 XM_017025303.1 NM_001130090.1 NM_031294.3 XM_006721583.3 XM_011524018.2 XM_011524019.2 XM_011524020.2 XM_011524021.2 XM_011524023.2 XM_011524025.2 XM_011524026.2

Gene Symbol
DRC3
Entrez Gene ID
83450
Gene Name
dynein regulatory complex subunit 3
Gene Aliases
CFAP134, LRRC48
UniGene
Hs.579264
Gene Chromosome Location
Chr.17: 17972766-18016889 on Build GRCh38
Transcripts
XM_011524065.1 XM_017025303.1 NM_001130090.1 NM_031294.3 XM_006721583.3 XM_011524018.2 XM_011524019.2 XM_011524020.2 XM_011524021.2 XM_011524023.2 XM_011524025.2 XM_011524026.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4649573 c.1437G>A p.W479* Chr.17: 18016174-18016174 on Build GRCh38
1256830 c.1348A>G p.I450V Chr.17: 18016085-18016085 on Build GRCh38
4699121 c.1362delC p.A456fs*8 Chr.17: 18016099-18016099 on Build GRCh38
4838961 c.1330T>C p.F444L Chr.17: 18016067-18016067 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs575558684 Chr.17: 18016132-18016132 on Build GRCh38
rs750647840 Chr.17: 18016099-18016099 on Build GRCh38
rs202171250 Chr.17: 18016112-18016112 on Build GRCh38
rs762367302 Chr.17: 18016181-18016181 on Build GRCh38
rs369426371 Chr.17: 18016017-18016017 on Build GRCh38
rs766485033 Chr.17: 18016192-18016192 on Build GRCh38
rs745798344 Chr.17: 18016089-18016089 on Build GRCh38
rs768670340 Chr.17: 18016041-18016041 on Build GRCh38
rs987621930 Chr.17: 18016236-18016236 on Build GRCh38
rs780351895 Chr.17: 18016105-18016105 on Build GRCh38
rs919046649 Chr.17: 18016208-18016208 on Build GRCh38
rs201011676 Chr.17: 18016059-18016059 on Build GRCh38
rs571427963 Chr.17: 18016073-18016073 on Build GRCh38
rs78556652 Chr.17: 18016083-18016083 on Build GRCh38
rs370315704 Chr.17: 18016108-18016108 on Build GRCh38
rs774076446 Chr.17: 18016140-18016140 on Build GRCh38
rs374340023 Chr.17: 18016079-18016079 on Build GRCh38
rs776729046 Chr.17: 18016042-18016042 on Build GRCh38
rs200381151 Chr.17: 18016100-18016100 on Build GRCh38
rs201748313 Chr.17: 18016049-18016049 on Build GRCh38
rs757590413 Chr.17: 18016085-18016085 on Build GRCh38
rs568795488 Chr.17: 18016058-18016058 on Build GRCh38
rs187171866 Chr.17: 18016084-18016084 on Build GRCh38
rs905125937 Chr.17: 18016134-18016134 on Build GRCh38
rs752836364 Chr.17: 18016244-18016244 on Build GRCh38
rs751869663 Chr.17: 18016216-18016216 on Build GRCh38
rs141679194 Chr.17: 18016071-18016071 on Build GRCh38
rs868267139 Chr.17: 18016155-18016155 on Build GRCh38
rs770452171 Chr.17: 18016139-18016139 on Build GRCh38
rs746121196 Chr.17: 18016019-18016019 on Build GRCh38
rs763394010 Chr.17: 18016189-18016189 on Build GRCh38
rs765775066 Chr.17: 18016182-18016182 on Build GRCh38
rs566959966 Chr.17: 18016032-18016032 on Build GRCh38
rs781477150 Chr.17: 18016234-18016234 on Build GRCh38
rs746343334 Chr.17: 18016021-18016023 on Build GRCh38
rs769696903 Chr.17: 18016126-18016126 on Build GRCh38
rs759026512 Chr.17: 18016161-18016161 on Build GRCh38
rs1009348214 Chr.17: 18016198-18016198 on Build GRCh38
rs78467765 Chr.17: 18016025-18016025 on Build GRCh38
rs973474997 Chr.17: 18016162-18016162 on Build GRCh38
rs569837531 Chr.17: 18016168-18016168 on Build GRCh38
rs748163277 Chr.17: 18016124-18016124 on Build GRCh38
rs757954586 Chr.17: 18016243-18016243 on Build GRCh38
rs919118825 Chr.17: 18016102-18016102 on Build GRCh38
rs762569885 Chr.17: 18016062-18016062 on Build GRCh38
rs750711539 Chr.17: 18016188-18016188 on Build GRCh38
rs375626870 Chr.17: 18016131-18016131 on Build GRCh38
rs755155187 Chr.17: 18016111-18016111 on Build GRCh38
rs755104187 Chr.17: 18016231-18016231 on Build GRCh38
rs371092039 Chr.17: 18016164-18016164 on Build GRCh38

View Full Product Details