Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 130341974-130342201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC115
Entrez Gene ID
84317
Gene Name
coiled-coil domain containing 115
Gene Aliases
CDG2O, ccp1
UniGene
Hs.104203
Gene Chromosome Location
Chr.2: 130337933-130342681 on Build GRCh38
Transcripts
NM_001321118.1 NM_001321119.1 NM_032357.3 NR_104471.1 NR_135548.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
279613 c.104T>C p.V35A Chr.2: 130342022-130342022 on Build GRCh38
6010276 c.75G>A p.K25K Chr.2: 130342155-130342155 on Build GRCh38
6621344 c.96C>T p.N32N Chr.2: 130342030-130342030 on Build GRCh38
255594 c.11T>C p.L4P Chr.2: 130342115-130342115 on Build GRCh38
6621342 c.3G>C p.M1I Chr.2: 130342123-130342123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374223206 Chr.2: 130342149-130342149 on Build GRCh38
rs1035273120 Chr.2: 130341997-130341998 on Build GRCh38
rs371413716 Chr.2: 130342146-130342146 on Build GRCh38
rs752443267 Chr.2: 130342171-130342171 on Build GRCh38
rs751429407 Chr.2: 130342113-130342113 on Build GRCh38
rs758351091 Chr.2: 130342030-130342030 on Build GRCh38
rs866944473 Chr.2: 130342162-130342163 on Build GRCh38
rs759432544 Chr.2: 130342115-130342115 on Build GRCh38
rs763799502 Chr.2: 130342147-130342147 on Build GRCh38
rs780222452 Chr.2: 130342153-130342153 on Build GRCh38
rs763766228 Chr.2: 130342010-130342010 on Build GRCh38
rs752919660 Chr.2: 130342125-130342125 on Build GRCh38
rs763011464 Chr.2: 130342110-130342110 on Build GRCh38
rs762600539 Chr.2: 130342159-130342159 on Build GRCh38
rs869025583 Chr.2: 130342095-130342095 on Build GRCh38
rs976330400 Chr.2: 130342079-130342079 on Build GRCh38
rs766377161 Chr.2: 130342112-130342112 on Build GRCh38
rs776120445 Chr.2: 130342011-130342011 on Build GRCh38
rs757297743 Chr.2: 130342142-130342142 on Build GRCh38
rs1003677034 Chr.2: 130341995-130341995 on Build GRCh38
rs1017292890 Chr.2: 130342066-130342066 on Build GRCh38
rs771291770 Chr.2: 130341996-130341996 on Build GRCh38
rs368236865 Chr.2: 130342165-130342165 on Build GRCh38
rs769526847 Chr.2: 130342156-130342156 on Build GRCh38
rs750387342 Chr.2: 130342023-130342023 on Build GRCh38
rs966809969 Chr.2: 130342116-130342116 on Build GRCh38
rs553740638 Chr.2: 130342048-130342048 on Build GRCh38
rs754697908 Chr.2: 130342039-130342039 on Build GRCh38
rs761461065 Chr.2: 130342018-130342018 on Build GRCh38
rs190933486 Chr.2: 130342170-130342170 on Build GRCh38
rs748303723 Chr.2: 130342045-130342045 on Build GRCh38
rs774721477 Chr.2: 130342001-130342001 on Build GRCh38
rs887570847 Chr.2: 130342036-130342036 on Build GRCh38
rs756251999 Chr.2: 130342055-130342055 on Build GRCh38
rs770576908 Chr.2: 130342116-130342117 on Build GRCh38
rs776405320 Chr.2: 130342091-130342091 on Build GRCh38
rs769438903 Chr.2: 130342100-130342100 on Build GRCh38
rs150332171 Chr.2: 130342033-130342033 on Build GRCh38
rs759324356 Chr.2: 130342166-130342166 on Build GRCh38
rs779177069 Chr.2: 130342077-130342077 on Build GRCh38
rs767209846 Chr.2: 130342121-130342121 on Build GRCh38
rs375455175 Chr.2: 130342158-130342158 on Build GRCh38
rs749812085 Chr.2: 130342065-130342065 on Build GRCh38
rs925361476 Chr.2: 130342127-130342127 on Build GRCh38
rs760919249 Chr.2: 130342174-130342174 on Build GRCh38
rs746287541 Chr.2: 130342078-130342078 on Build GRCh38
rs201206111 Chr.2: 130342070-130342070 on Build GRCh38
rs370742471 Chr.2: 130342060-130342060 on Build GRCh38
rs754040993 Chr.2: 130342179-130342179 on Build GRCh38
rs774189038 Chr.2: 130342128-130342129 on Build GRCh38
rs760268834 Chr.2: 130342008-130342008 on Build GRCh38
rs374624586 Chr.2: 130342107-130342107 on Build GRCh38
rs772557570 Chr.2: 130342150-130342150 on Build GRCh38
rs146081742 Chr.2: 130342099-130342099 on Build GRCh38
rs777854522 Chr.2: 130342128-130342128 on Build GRCh38
rs781526342 Chr.2: 130342044-130342044 on Build GRCh38
rs764341463 Chr.2: 130342177-130342177 on Build GRCh38
rs756272454 Chr.2: 130342126-130342126 on Build GRCh38
rs764828737 Chr.2: 130342019-130342019 on Build GRCh38
rs772306064 Chr.2: 130342090-130342090 on Build GRCh38
rs751325113 Chr.2: 130342034-130342034 on Build GRCh38
rs747823334 Chr.2: 130342155-130342155 on Build GRCh38
rs757419653 Chr.2: 130342181-130342181 on Build GRCh38
rs199963720 Chr.2: 130342141-130342141 on Build GRCh38

View Full Product Details