Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 93952901-93953166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BARX1
Entrez Gene ID
56033
Gene Name
BARX homeobox 1
Gene Aliases
-
UniGene
Hs.164960
Gene Chromosome Location
Chr.9: 93951627-93955326 on Build GRCh38
Transcripts
NM_021570.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1111312 c.414A>G p.K138K Chr.9: 93952997-93952997 on Build GRCh38
1184445 c.371G>A p.R124H Chr.9: 93953040-93953040 on Build GRCh38
6431461 c.310G>A p.A104T Chr.9: 93953101-93953101 on Build GRCh38
1463739 c.427C>T p.R143C Chr.9: 93952984-93952984 on Build GRCh38
5548340 c.308C>T p.A103V Chr.9: 93953103-93953103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774679647 Chr.9: 93953051-93953051 on Build GRCh38
rs767061973 Chr.9: 93953098-93953098 on Build GRCh38
rs761812989 Chr.9: 93953064-93953064 on Build GRCh38
rs146922051 Chr.9: 93952985-93952985 on Build GRCh38
rs756513295 Chr.9: 93953012-93953012 on Build GRCh38
rs772117798 Chr.9: 93953141-93953141 on Build GRCh38
rs544075964 Chr.9: 93953095-93953095 on Build GRCh38
rs1343899 Chr.9: 93953125-93953125 on Build GRCh38
rs748707341 Chr.9: 93953005-93953005 on Build GRCh38
rs900626317 Chr.9: 93953058-93953084 on Build GRCh38
rs562460428 Chr.9: 93953123-93953123 on Build GRCh38
rs752147918 Chr.9: 93953107-93953107 on Build GRCh38
rs770498173 Chr.9: 93953009-93953009 on Build GRCh38
rs774480658 Chr.9: 93953024-93953024 on Build GRCh38
rs751109837 Chr.9: 93952928-93952928 on Build GRCh38
rs778400548 Chr.9: 93953118-93953118 on Build GRCh38
rs780522907 Chr.9: 93953001-93953001 on Build GRCh38
rs866954519 Chr.9: 93952977-93952977 on Build GRCh38
rs565295556 Chr.9: 93952943-93952943 on Build GRCh38
rs746115723 Chr.9: 93953136-93953136 on Build GRCh38
rs541371621 Chr.9: 93953146-93953146 on Build GRCh38
rs201946673 Chr.9: 93953080-93953080 on Build GRCh38
rs369928804 Chr.9: 93953026-93953026 on Build GRCh38
rs549526966 Chr.9: 93953131-93953131 on Build GRCh38
rs771325755 Chr.9: 93953048-93953048 on Build GRCh38
rs1044700919 Chr.9: 93953077-93953077 on Build GRCh38
rs778351778 Chr.9: 93953016-93953016 on Build GRCh38
rs775374432 Chr.9: 93953063-93953063 on Build GRCh38
rs1013682813 Chr.9: 93953069-93953069 on Build GRCh38
rs773041333 Chr.9: 93953084-93953084 on Build GRCh38
rs780245526 Chr.9: 93953148-93953148 on Build GRCh38
rs1032506090 Chr.9: 93953093-93953093 on Build GRCh38
rs1047001899 Chr.9: 93953000-93953000 on Build GRCh38
rs756740714 Chr.9: 93953112-93953112 on Build GRCh38
rs868126121 Chr.9: 93952988-93952988 on Build GRCh38
rs373384950 Chr.9: 93953044-93953044 on Build GRCh38
rs754547962 Chr.9: 93952992-93952992 on Build GRCh38
rs868608877 Chr.9: 93953019-93953019 on Build GRCh38
rs542644678 Chr.9: 93953057-93953057 on Build GRCh38
rs1051042948 Chr.9: 93953021-93953021 on Build GRCh38
rs772214631 Chr.9: 93953062-93953062 on Build GRCh38
rs765995311 Chr.9: 93952926-93952926 on Build GRCh38
rs532945486 Chr.9: 93953127-93953127 on Build GRCh38
rs45527632 Chr.9: 93953104-93953104 on Build GRCh38
rs531602020 Chr.9: 93953092-93953092 on Build GRCh38

View Full Product Details