Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 116398881-116399134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1A1
Entrez Gene ID
476
Gene Name
ATPase Na+/K+ transporting subunit alpha 1
Gene Aliases
-
UniGene
Hs.371889
Gene Chromosome Location
Chr.1: 116373173-116404774 on Build GRCh38
Transcripts
NM_000701.7 NM_001160233.1 NM_001160234.1 XM_017001360.1 XM_017001361.1 NR_027645.1 NR_027646.1

Gene Symbol
ATP1A1-AS1
Entrez Gene ID
84852
Gene Name
ATP1A1 antisense RNA 1
Gene Aliases
ATP1A1OS, C1orf203
UniGene
Hs.664752
Gene Chromosome Location
Chr.1: 116392865-116418622 on Build GRCh38
Transcripts
NM_000701.7 NM_001160233.1 NM_001160234.1 XM_017001360.1 XM_017001361.1 NR_027645.1 NR_027646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6459175 c.2324C>T p.S775F Chr.1: 116398960-116398960 on Build GRCh38
1600607 c.2446A>G p.M816V Chr.1: 116399082-116399082 on Build GRCh38
4021027 c.2323T>C p.S775P Chr.1: 116398959-116398959 on Build GRCh38
3471876 c.2343C>T p.T781T Chr.1: 116398979-116398979 on Build GRCh38
1245704 c.2294G>A p.G765D Chr.1: 116398930-116398930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375568278 Chr.1: 116398943-116398943 on Build GRCh38
rs141378291 Chr.1: 116398961-116398961 on Build GRCh38
rs751282376 Chr.1: 116398935-116398935 on Build GRCh38
rs776767098 Chr.1: 116399022-116399022 on Build GRCh38
rs919214347 Chr.1: 116399027-116399027 on Build GRCh38
rs779747462 Chr.1: 116399112-116399112 on Build GRCh38
rs756962374 Chr.1: 116399097-116399097 on Build GRCh38
rs143154361 Chr.1: 116398934-116398934 on Build GRCh38
rs765164068 Chr.1: 116398909-116398909 on Build GRCh38
rs531429824 Chr.1: 116398915-116398915 on Build GRCh38
rs780771210 Chr.1: 116398915-116398915 on Build GRCh38
rs372034384 Chr.1: 116399048-116399048 on Build GRCh38
rs767286476 Chr.1: 116399098-116399098 on Build GRCh38
rs985958983 Chr.1: 116398985-116398985 on Build GRCh38
rs878888834 Chr.1: 116399089-116399089 on Build GRCh38
rs887503868 Chr.1: 116399103-116399103 on Build GRCh38
rs773672785 Chr.1: 116399000-116399000 on Build GRCh38
rs116412562 Chr.1: 116399109-116399109 on Build GRCh38
rs758253415 Chr.1: 116398917-116398917 on Build GRCh38
rs745407330 Chr.1: 116398964-116398964 on Build GRCh38
rs373372932 Chr.1: 116399110-116399110 on Build GRCh38
rs772154740 Chr.1: 116398906-116398906 on Build GRCh38
rs751394163 Chr.1: 116399095-116399095 on Build GRCh38
rs1003114854 Chr.1: 116398910-116398910 on Build GRCh38
rs200315945 Chr.1: 116399003-116399003 on Build GRCh38
rs748750456 Chr.1: 116398988-116398988 on Build GRCh38
rs878964637 Chr.1: 116399088-116399088 on Build GRCh38
rs763917443 Chr.1: 116399087-116399087 on Build GRCh38
rs747283959 Chr.1: 116399002-116399002 on Build GRCh38
rs111483355 Chr.1: 116399075-116399075 on Build GRCh38
rs1031963587 Chr.1: 116398925-116398925 on Build GRCh38
rs530130545 Chr.1: 116398967-116398967 on Build GRCh38
rs956023678 Chr.1: 116398933-116398933 on Build GRCh38
rs754256658 Chr.1: 116398914-116398914 on Build GRCh38
rs377635301 Chr.1: 116398926-116398926 on Build GRCh38
rs144039896 Chr.1: 116398991-116398991 on Build GRCh38
rs759625415 Chr.1: 116399045-116399045 on Build GRCh38
rs775541230 Chr.1: 116399063-116399063 on Build GRCh38
rs944570753 Chr.1: 116398997-116398997 on Build GRCh38
rs1049210174 Chr.1: 116399086-116399086 on Build GRCh38
rs779579313 Chr.1: 116398979-116398979 on Build GRCh38

View Full Product Details