Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150183635-150183908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDX1
Entrez Gene ID
1044
Gene Name
caudal type homeobox 1
Gene Aliases
-
UniGene
Hs.1545
Gene Chromosome Location
Chr.5: 150166781-150184558 on Build GRCh38
Transcripts
NM_001804.2

SNP Detail

SNP ID Chromosome Location
rs1054751648 Chr.5: 150183864-150183864 on Build GRCh38
rs748673533 Chr.5: 150183711-150183711 on Build GRCh38
rs980951066 Chr.5: 150183820-150183820 on Build GRCh38
rs753762260 Chr.5: 150183683-150183683 on Build GRCh38
rs939235573 Chr.5: 150183850-150183850 on Build GRCh38
rs781693708 Chr.5: 150183703-150183703 on Build GRCh38
rs547789728 Chr.5: 150183768-150183768 on Build GRCh38
rs990374450 Chr.5: 150183805-150183805 on Build GRCh38
rs556736390 Chr.5: 150183662-150183662 on Build GRCh38
rs766624791 Chr.5: 150183702-150183702 on Build GRCh38
rs939198500 Chr.5: 150183707-150183708 on Build GRCh38
rs765147524 Chr.5: 150183694-150183694 on Build GRCh38
rs749716046 Chr.5: 150183719-150183719 on Build GRCh38
rs778111175 Chr.5: 150183718-150183718 on Build GRCh38
rs575862167 Chr.5: 150183835-150183835 on Build GRCh38
rs745932017 Chr.5: 150183732-150183732 on Build GRCh38
rs899309371 Chr.5: 150183736-150183736 on Build GRCh38
rs964825479 Chr.5: 150183781-150183781 on Build GRCh38
rs1026997246 Chr.5: 150183750-150183750 on Build GRCh38
rs769806977 Chr.5: 150183760-150183760 on Build GRCh38
rs771176011 Chr.5: 150183720-150183720 on Build GRCh38
rs939065697 Chr.5: 150183886-150183886 on Build GRCh38
rs764073764 Chr.5: 150183658-150183658 on Build GRCh38
rs775264282 Chr.5: 150183807-150183807 on Build GRCh38
rs2880319 Chr.5: 150183762-150183762 on Build GRCh38
rs770083616 Chr.5: 150183715-150183715 on Build GRCh38
rs1017583536 Chr.5: 150183778-150183778 on Build GRCh38
rs539226569 Chr.5: 150183722-150183722 on Build GRCh38
rs989153544 Chr.5: 150183684-150183684 on Build GRCh38
rs927885014 Chr.5: 150183844-150183844 on Build GRCh38
rs966935252 Chr.5: 150183798-150183798 on Build GRCh38
rs745329598 Chr.5: 150183737-150183737 on Build GRCh38
rs145345944 Chr.5: 150183700-150183700 on Build GRCh38
rs1008905630 Chr.5: 150183775-150183775 on Build GRCh38
rs201588716 Chr.5: 150183653-150183653 on Build GRCh38
rs796758451 Chr.5: 150183783-150183783 on Build GRCh38
rs954919997 Chr.5: 150183766-150183766 on Build GRCh38
rs915104990 Chr.5: 150183792-150183792 on Build GRCh38
rs969066931 Chr.5: 150183814-150183814 on Build GRCh38
rs535027006 Chr.5: 150183699-150183699 on Build GRCh38
rs955768406 Chr.5: 150183763-150183763 on Build GRCh38
rs187114829 Chr.5: 150183701-150183701 on Build GRCh38
rs369619598 Chr.5: 150183689-150183689 on Build GRCh38
rs1042680588 Chr.5: 150183660-150183660 on Build GRCh38
rs956391871 Chr.5: 150183675-150183675 on Build GRCh38
rs557920185 Chr.5: 150183764-150183764 on Build GRCh38
rs753962349 Chr.5: 150183755-150183755 on Build GRCh38
rs201901105 Chr.5: 150183696-150183696 on Build GRCh38
rs574983359 Chr.5: 150183870-150183870 on Build GRCh38

View Full Product Details