Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 83280068-83280327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC90B
Entrez Gene ID
60492
Gene Name
coiled-coil domain containing 90B
Gene Aliases
MDS011, MDS025
UniGene
Hs.368866
Gene Chromosome Location
Chr.11: 83259093-83286407 on Build GRCh38
Transcripts
NM_001286116.1 NM_001286117.1 NM_001286118.1 NM_001286119.1 NM_001286120.1 NM_021825.4 XM_005274154.2 XM_005274156.3 XM_005274159.3 XM_017018103.1 XM_017018104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1739783 c.218A>T p.H73L Chr.11: 83280143-83280143 on Build GRCh38
3700156 c.175T>G p.L59V Chr.11: 83280186-83280186 on Build GRCh38
1605296 c.173A>T p.K58I Chr.11: 83280188-83280188 on Build GRCh38
3453953 c.163G>A p.E55K Chr.11: 83280198-83280198 on Build GRCh38
2220735 c.193G>A p.A65T Chr.11: 83280168-83280168 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763647703 Chr.11: 83280258-83280258 on Build GRCh38
rs749481342 Chr.11: 83280178-83280179 on Build GRCh38
rs776529576 Chr.11: 83280239-83280239 on Build GRCh38
rs756244696 Chr.11: 83280124-83280131 on Build GRCh38
rs267603220 Chr.11: 83280203-83280203 on Build GRCh38
rs766901640 Chr.11: 83280275-83280275 on Build GRCh38
rs780376823 Chr.11: 83280191-83280191 on Build GRCh38
rs777779300 Chr.11: 83280225-83280225 on Build GRCh38
rs771211764 Chr.11: 83280276-83280277 on Build GRCh38
rs759235734 Chr.11: 83280143-83280143 on Build GRCh38
rs772193955 Chr.11: 83280222-83280222 on Build GRCh38
rs762579826 Chr.11: 83280252-83280252 on Build GRCh38
rs745332815 Chr.11: 83280102-83280102 on Build GRCh38
rs752103195 Chr.11: 83280169-83280169 on Build GRCh38
rs375397191 Chr.11: 83280119-83280119 on Build GRCh38
rs777764921 Chr.11: 83280160-83280184 on Build GRCh38
rs201485542 Chr.11: 83280170-83280170 on Build GRCh38
rs372541981 Chr.11: 83280291-83280291 on Build GRCh38
rs368318377 Chr.11: 83280240-83280240 on Build GRCh38
rs755348738 Chr.11: 83280279-83280279 on Build GRCh38
rs771033874 Chr.11: 83280233-83280233 on Build GRCh38
rs141993989 Chr.11: 83280205-83280205 on Build GRCh38
rs769276895 Chr.11: 83280243-83280243 on Build GRCh38
rs761138038 Chr.11: 83280274-83280274 on Build GRCh38
rs776623523 Chr.11: 83280123-83280123 on Build GRCh38
rs765016606 Chr.11: 83280154-83280154 on Build GRCh38
rs777589186 Chr.11: 83280120-83280120 on Build GRCh38
rs754239235 Chr.11: 83280193-83280193 on Build GRCh38
rs1041698841 Chr.11: 83280245-83280245 on Build GRCh38
rs755152154 Chr.11: 83280200-83280200 on Build GRCh38
rs760476781 Chr.11: 83280112-83280112 on Build GRCh38
rs746926847 Chr.11: 83280226-83280226 on Build GRCh38
rs752956016 Chr.11: 83280288-83280288 on Build GRCh38
rs548508996 Chr.11: 83280281-83280281 on Build GRCh38
rs61747435 Chr.11: 83280221-83280221 on Build GRCh38
rs767845923 Chr.11: 83280174-83280174 on Build GRCh38
rs775212329 Chr.11: 83280250-83280250 on Build GRCh38
rs754151475 Chr.11: 83280277-83280277 on Build GRCh38
rs752683491 Chr.11: 83280105-83280105 on Build GRCh38
rs759429920 Chr.11: 83280242-83280242 on Build GRCh38
rs750937198 Chr.11: 83280185-83280185 on Build GRCh38
rs201496466 Chr.11: 83280190-83280190 on Build GRCh38
rs202008487 Chr.11: 83280121-83280121 on Build GRCh38
rs751120215 Chr.11: 83280273-83280273 on Build GRCh38
rs777884263 Chr.11: 83280293-83280293 on Build GRCh38

View Full Product Details