Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 108868753-108869026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC2
Entrez Gene ID
84071
Gene Name
armadillo repeat containing 2
Gene Aliases
bA787I22.1
UniGene
Hs.645481
Gene Chromosome Location
Chr.6: 108848410-108990738 on Build GRCh38
Transcripts
NM_001286609.1 NM_032131.5 XM_005267154.3 XM_005267155.2 XM_005267157.3 XM_006715573.2 XM_006715574.2 XM_011536166.1 XM_011536167.1 XM_011536168.2 XM_011536170.2 XM_011536172.2 XM_011536176.2 XM_017011350.1 XM_017011351.1 XM_017011352.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7287261 c.424C>T p.R142W Chr.6: 108868956-108868956 on Build GRCh38
1732827 c.410G>A p.R137K Chr.6: 108868942-108868942 on Build GRCh38
4901592 c.333C>T p.S111S Chr.6: 108868865-108868865 on Build GRCh38
1071916 c.401G>A p.R134H Chr.6: 108868933-108868933 on Build GRCh38
4910800 c.313C>G p.P105A Chr.6: 108868845-108868845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568064107 Chr.6: 108868978-108868978 on Build GRCh38
rs373388431 Chr.6: 108868782-108868782 on Build GRCh38
rs201194523 Chr.6: 108868803-108868803 on Build GRCh38
rs748227632 Chr.6: 108868788-108868788 on Build GRCh38
rs184241545 Chr.6: 108868783-108868783 on Build GRCh38
rs373132834 Chr.6: 108868929-108868929 on Build GRCh38
rs776082535 Chr.6: 108868863-108868863 on Build GRCh38
rs372533595 Chr.6: 108868900-108868900 on Build GRCh38
rs761425113 Chr.6: 108868883-108868883 on Build GRCh38
rs753358737 Chr.6: 108868878-108868878 on Build GRCh38
rs774041490 Chr.6: 108868805-108868806 on Build GRCh38
rs761127076 Chr.6: 108868941-108868941 on Build GRCh38
rs781134438 Chr.6: 108868988-108868988 on Build GRCh38
rs756932457 Chr.6: 108868781-108868781 on Build GRCh38
rs200798300 Chr.6: 108868987-108868987 on Build GRCh38
rs114398565 Chr.6: 108868784-108868784 on Build GRCh38
rs777752630 Chr.6: 108868918-108868918 on Build GRCh38
rs753492918 Chr.6: 108868776-108868776 on Build GRCh38
rs774902629 Chr.6: 108868836-108868836 on Build GRCh38
rs151269619 Chr.6: 108868864-108868864 on Build GRCh38
rs747690401 Chr.6: 108868967-108868968 on Build GRCh38
rs768397124 Chr.6: 108868796-108868796 on Build GRCh38
rs138675941 Chr.6: 108868955-108868955 on Build GRCh38
rs539620053 Chr.6: 108868969-108868969 on Build GRCh38
rs749537185 Chr.6: 108868828-108868828 on Build GRCh38
rs369716237 Chr.6: 108868922-108868922 on Build GRCh38
rs141810675 Chr.6: 108868991-108868991 on Build GRCh38
rs917788344 Chr.6: 108868906-108868906 on Build GRCh38
rs754936975 Chr.6: 108868907-108868907 on Build GRCh38
rs901412556 Chr.6: 108868979-108868979 on Build GRCh38
rs897499251 Chr.6: 108868799-108868799 on Build GRCh38
rs753893918 Chr.6: 108868990-108868990 on Build GRCh38
rs199686045 Chr.6: 108868891-108868891 on Build GRCh38
rs373107813 Chr.6: 108868956-108868956 on Build GRCh38
rs73524086 Chr.6: 108868829-108868829 on Build GRCh38
rs144751024 Chr.6: 108868937-108868937 on Build GRCh38
rs1002401383 Chr.6: 108868846-108868846 on Build GRCh38
rs143251061 Chr.6: 108868857-108868857 on Build GRCh38
rs759930999 Chr.6: 108868841-108868841 on Build GRCh38
rs554204061 Chr.6: 108868957-108868957 on Build GRCh38
rs1015707475 Chr.6: 108868822-108868822 on Build GRCh38
rs377502169 Chr.6: 108868933-108868933 on Build GRCh38
rs751335272 Chr.6: 108868986-108868986 on Build GRCh38
rs1027420970 Chr.6: 108868819-108868819 on Build GRCh38
rs199572050 Chr.6: 108868992-108868992 on Build GRCh38
rs750016179 Chr.6: 108868962-108868962 on Build GRCh38
rs763579807 Chr.6: 108868869-108868869 on Build GRCh38
rs77789034 Chr.6: 108868812-108868812 on Build GRCh38
rs755787855 Chr.6: 108868970-108868970 on Build GRCh38
rs750253894 Chr.6: 108868890-108868890 on Build GRCh38
rs140505039 Chr.6: 108868923-108868923 on Build GRCh38
rs764897142 Chr.6: 108868888-108868888 on Build GRCh38
rs571006027 Chr.6: 108868961-108868961 on Build GRCh38

View Full Product Details