Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35777573-35777800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP33
Entrez Gene ID
115703
Gene Name
Rho GTPase activating protein 33
Gene Aliases
NOMA-GAP, SNX26, TCGAP
UniGene
Hs.515364
Gene Chromosome Location
Chr.19: 35775515-35788822 on Build GRCh38
Transcripts
NM_001172630.1 NM_052948.3 XM_005258488.1 XM_006722999.1 XM_011526416.1 XM_011526417.1 XM_011526418.1 XM_011526419.1 XM_011526420.1 XM_011526421.2 XM_017026238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7297822 c.65G>A p.R22H Chr.19: 35777649-35777649 on Build GRCh38
1589996 c.30C>A p.L10L Chr.19: 35777614-35777614 on Build GRCh38
1589997 c.17T>G p.L6R Chr.19: 35777601-35777601 on Build GRCh38
6214728 c.68G>C p.S23T Chr.19: 35777652-35777652 on Build GRCh38
5680604 c.89C>T p.P30L Chr.19: 35777673-35777673 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754807634 Chr.19: 35777708-35777708 on Build GRCh38
rs537032920 Chr.19: 35777745-35777745 on Build GRCh38
rs749023544 Chr.19: 35777601-35777601 on Build GRCh38
rs750614808 Chr.19: 35777664-35777664 on Build GRCh38
rs373444720 Chr.19: 35777655-35777655 on Build GRCh38
rs199905706 Chr.19: 35777696-35777696 on Build GRCh38
rs775808859 Chr.19: 35777762-35777762 on Build GRCh38
rs761814922 Chr.19: 35777775-35777775 on Build GRCh38
rs779705107 Chr.19: 35777684-35777684 on Build GRCh38
rs764537904 Chr.19: 35777768-35777768 on Build GRCh38
rs757351355 Chr.19: 35777648-35777648 on Build GRCh38
rs756430555 Chr.19: 35777681-35777681 on Build GRCh38
rs772849980 Chr.19: 35777714-35777714 on Build GRCh38
rs368825074 Chr.19: 35777649-35777649 on Build GRCh38
rs915044601 Chr.19: 35777703-35777703 on Build GRCh38
rs774853963 Chr.19: 35777771-35777771 on Build GRCh38
rs1024049483 Chr.19: 35777748-35777748 on Build GRCh38
rs751605058 Chr.19: 35777598-35777598 on Build GRCh38
rs763346567 Chr.19: 35777765-35777765 on Build GRCh38
rs946607857 Chr.19: 35777732-35777732 on Build GRCh38
rs771492571 Chr.19: 35777713-35777713 on Build GRCh38
rs867349554 Chr.19: 35777733-35777733 on Build GRCh38
rs199692242 Chr.19: 35777685-35777685 on Build GRCh38
rs767491676 Chr.19: 35777780-35777780 on Build GRCh38
rs747609548 Chr.19: 35777705-35777705 on Build GRCh38
rs754936865 Chr.19: 35777687-35777687 on Build GRCh38
rs749147548 Chr.19: 35777686-35777686 on Build GRCh38
rs906912111 Chr.19: 35777751-35777751 on Build GRCh38
rs1054648255 Chr.19: 35777676-35777676 on Build GRCh38
rs1035030423 Chr.19: 35777679-35777679 on Build GRCh38
rs770673568 Chr.19: 35777755-35777755 on Build GRCh38
rs867667942 Chr.19: 35777636-35777636 on Build GRCh38

View Full Product Details