Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 56961921-56962162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CETP
Entrez Gene ID
1071
Gene Name
cholesteryl ester transfer protein
Gene Aliases
BPIFF, HDLCQ10
UniGene
Hs.89538
Gene Chromosome Location
Chr.16: 56961923-56983844 on Build GRCh38
Transcripts
NM_000078.2 NM_001286085.1 XM_006721124.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4671836 c.105T>C p.P35P Chr.16: 56962084-56962084 on Build GRCh38
2836109 c.51C>T p.A17A Chr.16: 56962030-56962030 on Build GRCh38
4061361 c.11C>T p.A4V Chr.16: 56961990-56961990 on Build GRCh38
1742997 c.73G>A p.E25K Chr.16: 56962052-56962052 on Build GRCh38
3957662 c.58A>G p.K20E Chr.16: 56962037-56962037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761121093 Chr.16: 56961967-56961967 on Build GRCh38
rs748183484 Chr.16: 56961953-56961953 on Build GRCh38
rs747564264 Chr.16: 56962064-56962064 on Build GRCh38
rs34065661 Chr.16: 56962023-56962023 on Build GRCh38
rs773933829 Chr.16: 56962088-56962088 on Build GRCh38
rs780219684 Chr.16: 56961947-56961947 on Build GRCh38
rs755268936 Chr.16: 56962125-56962126 on Build GRCh38
rs771585518 Chr.16: 56962068-56962068 on Build GRCh38
rs750401042 Chr.16: 56962039-56962039 on Build GRCh38
rs772113814 Chr.16: 56962061-56962061 on Build GRCh38
rs1035637474 Chr.16: 56961980-56961980 on Build GRCh38
rs754790061 Chr.16: 56962120-56962120 on Build GRCh38
rs147758502 Chr.16: 56962071-56962071 on Build GRCh38
rs768295149 Chr.16: 56962084-56962084 on Build GRCh38
rs762643341 Chr.16: 56962009-56962009 on Build GRCh38
rs759178413 Chr.16: 56962119-56962119 on Build GRCh38
rs34119551 Chr.16: 56961996-56961996 on Build GRCh38
rs991188482 Chr.16: 56962002-56962002 on Build GRCh38
rs753665779 Chr.16: 56962048-56962048 on Build GRCh38
rs1002690375 Chr.16: 56961960-56961960 on Build GRCh38
rs529396716 Chr.16: 56962121-56962121 on Build GRCh38
rs780994834 Chr.16: 56961940-56961940 on Build GRCh38
rs767475225 Chr.16: 56962027-56962027 on Build GRCh38
rs142117489 Chr.16: 56962063-56962063 on Build GRCh38
rs914182923 Chr.16: 56962010-56962010 on Build GRCh38
rs773654189 Chr.16: 56961961-56961961 on Build GRCh38
rs5884 Chr.16: 56962045-56962045 on Build GRCh38
rs766825140 Chr.16: 56961991-56961992 on Build GRCh38
rs749958925 Chr.16: 56962096-56962096 on Build GRCh38
rs766986850 Chr.16: 56962094-56962094 on Build GRCh38
rs995506552 Chr.16: 56962132-56962132 on Build GRCh38
rs368028559 Chr.16: 56962130-56962130 on Build GRCh38
rs749561045 Chr.16: 56961949-56961949 on Build GRCh38
rs776131795 Chr.16: 56962098-56962098 on Build GRCh38
rs146270725 Chr.16: 56961993-56961993 on Build GRCh38
rs201234837 Chr.16: 56962052-56962052 on Build GRCh38
rs574035014 Chr.16: 56962047-56962047 on Build GRCh38
rs377094474 Chr.16: 56961944-56961944 on Build GRCh38
rs751916721 Chr.16: 56962098-56962101 on Build GRCh38
rs760567825 Chr.16: 56962101-56962101 on Build GRCh38
rs763683938 Chr.16: 56962021-56962021 on Build GRCh38
rs763366096 Chr.16: 56961972-56961972 on Build GRCh38
rs755959729 Chr.16: 56961945-56961945 on Build GRCh38
rs200386961 Chr.16: 56962033-56962033 on Build GRCh38
rs148628525 Chr.16: 56962051-56962051 on Build GRCh38
rs777093455 Chr.16: 56962070-56962070 on Build GRCh38
rs772575778 Chr.16: 56961958-56961958 on Build GRCh38
rs766310314 Chr.16: 56962111-56962111 on Build GRCh38
rs370507501 Chr.16: 56961950-56961950 on Build GRCh38
rs765808810 Chr.16: 56962092-56962093 on Build GRCh38
rs761381765 Chr.16: 56962092-56962092 on Build GRCh38

View Full Product Details