Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82956974-82957239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNTL1
Entrez Gene ID
146712
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase-like 1
Gene Aliases
3-Gn-T8, B3GNT8, BGnT-8, beta-1, beta3Gn-T8, beta3GnTL1
UniGene
Hs.607824
Gene Chromosome Location
Chr.17: 82942149-83051845 on Build GRCh38
Transcripts
NM_001009905.2 NM_001320742.1 NM_001320743.1 NR_135465.1 NR_135466.1 XM_006722272.3 XM_011523532.2 XM_011523535.1 XM_017024205.1 XM_017024206.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4187857 c.886G>T p.E296* Chr.17: 82957225-82957225 on Build GRCh38
3523921 c.916C>T p.H306Y Chr.17: 82957195-82957195 on Build GRCh38
6609370 c.885C>T p.D295D Chr.17: 82957226-82957226 on Build GRCh38
473585 c.895A>C p.I299L Chr.17: 82957216-82957216 on Build GRCh38
4070971 c.880G>A p.V294M Chr.17: 82957231-82957231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001040948 Chr.17: 82957020-82957020 on Build GRCh38
rs763147451 Chr.17: 82957192-82957192 on Build GRCh38
rs765292078 Chr.17: 82957180-82957180 on Build GRCh38
rs189922999 Chr.17: 82957141-82957141 on Build GRCh38
rs552876608 Chr.17: 82957010-82957010 on Build GRCh38
rs576139413 Chr.17: 82957081-82957081 on Build GRCh38
rs765431143 Chr.17: 82957134-82957134 on Build GRCh38
rs748985881 Chr.17: 82957145-82957145 on Build GRCh38
rs779646338 Chr.17: 82957144-82957144 on Build GRCh38
rs1001865 Chr.17: 82957112-82957112 on Build GRCh38
rs116508743 Chr.17: 82957110-82957110 on Build GRCh38
rs775442266 Chr.17: 82957188-82957188 on Build GRCh38
rs780417031 Chr.17: 82957088-82957088 on Build GRCh38
rs779386785 Chr.17: 82957007-82957007 on Build GRCh38
rs776948294 Chr.17: 82957179-82957179 on Build GRCh38
rs1531281 Chr.17: 82957002-82957002 on Build GRCh38
rs925281235 Chr.17: 82957161-82957161 on Build GRCh38
rs772644131 Chr.17: 82957206-82957206 on Build GRCh38
rs754154560 Chr.17: 82957132-82957132 on Build GRCh38
rs866440563 Chr.17: 82957157-82957157 on Build GRCh38
rs60745564 Chr.17: 82957033-82957033 on Build GRCh38
rs764500498 Chr.17: 82957193-82957193 on Build GRCh38
rs767889532 Chr.17: 82957219-82957219 on Build GRCh38
rs962540664 Chr.17: 82957120-82957120 on Build GRCh38
rs969134013 Chr.17: 82957078-82957078 on Build GRCh38
rs1052672796 Chr.17: 82957021-82957021 on Build GRCh38
rs778315762 Chr.17: 82957154-82957154 on Build GRCh38
rs764460973 Chr.17: 82957139-82957139 on Build GRCh38
rs774480625 Chr.17: 82957025-82957025 on Build GRCh38
rs375907083 Chr.17: 82957150-82957150 on Build GRCh38
rs1000595096 Chr.17: 82957086-82957086 on Build GRCh38
rs748063639 Chr.17: 82957156-82957156 on Build GRCh38
rs541674543 Chr.17: 82957133-82957133 on Build GRCh38
rs145692272 Chr.17: 82957065-82957065 on Build GRCh38
rs958762074 Chr.17: 82957108-82957108 on Build GRCh38
rs766708893 Chr.17: 82957130-82957130 on Build GRCh38
rs752148465 Chr.17: 82957200-82957200 on Build GRCh38
rs371262181 Chr.17: 82957217-82957217 on Build GRCh38
rs1013779601 Chr.17: 82957057-82957057 on Build GRCh38
rs370441124 Chr.17: 82957169-82957169 on Build GRCh38
rs767806412 Chr.17: 82957011-82957011 on Build GRCh38
rs201590747 Chr.17: 82957168-82957168 on Build GRCh38
rs753209606 Chr.17: 82957138-82957138 on Build GRCh38
rs1002377815 Chr.17: 82957079-82957079 on Build GRCh38
rs781624617 Chr.17: 82957183-82957184 on Build GRCh38
rs543405325 Chr.17: 82957104-82957104 on Build GRCh38
rs770902809 Chr.17: 82957176-82957176 on Build GRCh38
rs965574334 Chr.17: 82957111-82957111 on Build GRCh38
rs557786096 Chr.17: 82957080-82957080 on Build GRCh38
rs746769380 Chr.17: 82957173-82957173 on Build GRCh38

View Full Product Details