Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 101429748-101430001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDOB
Entrez Gene ID
229
Gene Name
aldolase, fructose-bisphosphate B
Gene Aliases
ALDB, ALDO2
UniGene
Hs.530274
Gene Chromosome Location
Chr.9: 101420560-101435780 on Build GRCh38
Transcripts
NM_000035.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1701426 c.304G>A p.G102R Chr.9: 101429775-101429775 on Build GRCh38
1701427 c.272G>A p.G91E Chr.9: 101429807-101429807 on Build GRCh38
6054989 c.243C>A p.H81Q Chr.9: 101429836-101429836 on Build GRCh38
4661074 c.178C>T p.R60* Chr.9: 101429901-101429901 on Build GRCh38
1701428 c.148G>A p.E50K Chr.9: 101429931-101429931 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772436722 Chr.9: 101429883-101429883 on Build GRCh38
rs372738117 Chr.9: 101429791-101429791 on Build GRCh38
rs867022678 Chr.9: 101429781-101429781 on Build GRCh38
rs777794664 Chr.9: 101429932-101429932 on Build GRCh38
rs1013015077 Chr.9: 101429944-101429944 on Build GRCh38
rs200050226 Chr.9: 101429912-101429912 on Build GRCh38
rs200121991 Chr.9: 101429786-101429786 on Build GRCh38
rs777050401 Chr.9: 101429904-101429904 on Build GRCh38
rs746149471 Chr.9: 101429877-101429877 on Build GRCh38
rs752642604 Chr.9: 101429919-101429919 on Build GRCh38
rs149671008 Chr.9: 101429923-101429923 on Build GRCh38
rs764779161 Chr.9: 101429852-101429852 on Build GRCh38
rs752633158 Chr.9: 101429870-101429870 on Build GRCh38
rs375679562 Chr.9: 101429855-101429855 on Build GRCh38
rs773510589 Chr.9: 101429794-101429794 on Build GRCh38
rs781126536 Chr.9: 101429784-101429784 on Build GRCh38
rs786204598 Chr.9: 101429964-101429967 on Build GRCh38
rs41281039 Chr.9: 101429943-101429943 on Build GRCh38
rs182003715 Chr.9: 101429836-101429836 on Build GRCh38
rs77849749 Chr.9: 101429960-101429960 on Build GRCh38
rs141267935 Chr.9: 101429900-101429900 on Build GRCh38
rs755336557 Chr.9: 101429958-101429958 on Build GRCh38
rs568367333 Chr.9: 101429831-101429831 on Build GRCh38
rs971472548 Chr.9: 101429979-101429979 on Build GRCh38
rs761063847 Chr.9: 101429824-101429824 on Build GRCh38
rs780442649 Chr.9: 101429951-101429951 on Build GRCh38
rs757318986 Chr.9: 101429871-101429871 on Build GRCh38
rs773642713 Chr.9: 101429976-101429976 on Build GRCh38
rs747604233 Chr.9: 101429856-101429856 on Build GRCh38
rs779129856 Chr.9: 101429872-101429872 on Build GRCh38
rs199920124 Chr.9: 101429942-101429942 on Build GRCh38
rs570143313 Chr.9: 101429910-101429910 on Build GRCh38
rs779797602 Chr.9: 101429769-101429769 on Build GRCh38
rs772473508 Chr.9: 101429952-101429952 on Build GRCh38
rs370761101 Chr.9: 101429909-101429909 on Build GRCh38
rs781023784 Chr.9: 101429858-101429858 on Build GRCh38
rs974582114 Chr.9: 101429777-101429777 on Build GRCh38
rs747019279 Chr.9: 101429770-101429770 on Build GRCh38
rs118204429 Chr.9: 101429901-101429901 on Build GRCh38
rs151219186 Chr.9: 101429888-101429888 on Build GRCh38
rs754861661 Chr.9: 101429782-101429782 on Build GRCh38
rs771004498 Chr.9: 101429790-101429790 on Build GRCh38
rs962536647 Chr.9: 101429776-101429776 on Build GRCh38
rs754735910 Chr.9: 101429857-101429857 on Build GRCh38
rs200585150 Chr.9: 101429815-101429815 on Build GRCh38
rs749333982 Chr.9: 101429788-101429788 on Build GRCh38
rs549682194 Chr.9: 101429797-101429797 on Build GRCh38
rs201397971 Chr.9: 101429913-101429913 on Build GRCh38
rs758508874 Chr.9: 101429945-101429945 on Build GRCh38
rs777153196 Chr.9: 101429835-101429835 on Build GRCh38
rs748663340 Chr.9: 101429967-101429967 on Build GRCh38
rs769043841 Chr.9: 101429829-101429829 on Build GRCh38
rs759204107 Chr.9: 101429853-101429853 on Build GRCh38
rs780352710 Chr.9: 101429892-101429892 on Build GRCh38
rs2854709 Chr.9: 101429917-101429917 on Build GRCh38
rs759273480 Chr.9: 101429914-101429914 on Build GRCh38

View Full Product Details