Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40163611-40163836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASC3
Entrez Gene ID
22794
Gene Name
cancer susceptibility candidate 3
Gene Aliases
BTZ, MLN51
UniGene
Hs.743287
Gene Chromosome Location
Chr.17: 40140254-40172178 on Build GRCh38
Transcripts
NM_007359.4 XM_005257163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1750008 c.1021G>A p.E341K Chr.17: 40163716-40163716 on Build GRCh38
6146643 c.1014G>T p.R338R Chr.17: 40163709-40163709 on Build GRCh38
3889618 c.1030A>T p.K344* Chr.17: 40163725-40163725 on Build GRCh38
6556348 c.1080G>T p.R360S Chr.17: 40163775-40163775 on Build GRCh38
6620366 c.916C>T p.R306C Chr.17: 40163611-40163611 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374144661 Chr.17: 40163776-40163776 on Build GRCh38
rs150678207 Chr.17: 40163690-40163690 on Build GRCh38
rs148407288 Chr.17: 40163727-40163727 on Build GRCh38
rs570391449 Chr.17: 40163781-40163781 on Build GRCh38
rs747367731 Chr.17: 40163775-40163775 on Build GRCh38
rs746420446 Chr.17: 40163660-40163674 on Build GRCh38
rs778134722 Chr.17: 40163766-40163766 on Build GRCh38
rs749510463 Chr.17: 40163638-40163638 on Build GRCh38
rs772561168 Chr.17: 40163709-40163709 on Build GRCh38
rs764903865 Chr.17: 40163663-40163663 on Build GRCh38
rs781021733 Chr.17: 40163749-40163749 on Build GRCh38
rs768958625 Chr.17: 40163701-40163701 on Build GRCh38
rs372980819 Chr.17: 40163639-40163639 on Build GRCh38
rs149859405 Chr.17: 40163714-40163714 on Build GRCh38
rs758162778 Chr.17: 40163671-40163671 on Build GRCh38
rs147487074 Chr.17: 40163677-40163677 on Build GRCh38
rs146454320 Chr.17: 40163754-40163754 on Build GRCh38
rs762300441 Chr.17: 40163647-40163647 on Build GRCh38
rs200515784 Chr.17: 40163648-40163648 on Build GRCh38
rs776169377 Chr.17: 40163721-40163721 on Build GRCh38
rs139973585 Chr.17: 40163708-40163708 on Build GRCh38
rs761153646 Chr.17: 40163654-40163654 on Build GRCh38
rs144021374 Chr.17: 40163743-40163743 on Build GRCh38
rs755077795 Chr.17: 40163753-40163753 on Build GRCh38
rs757149919 Chr.17: 40163747-40163747 on Build GRCh38
rs765901749 Chr.17: 40163655-40163655 on Build GRCh38
rs199653330 Chr.17: 40163683-40163683 on Build GRCh38
rs752445327 Chr.17: 40163668-40163668 on Build GRCh38
rs200624336 Chr.17: 40163720-40163720 on Build GRCh38
rs771349388 Chr.17: 40163715-40163715 on Build GRCh38
rs775035888 Chr.17: 40163802-40163802 on Build GRCh38
rs143077699 Chr.17: 40163662-40163662 on Build GRCh38
rs762658127 Chr.17: 40163742-40163742 on Build GRCh38
rs1055062596 Chr.17: 40163733-40163733 on Build GRCh38
rs751323290 Chr.17: 40163744-40163744 on Build GRCh38
rs776986603 Chr.17: 40163777-40163777 on Build GRCh38
rs201645449 Chr.17: 40163707-40163707 on Build GRCh38
rs138442252 Chr.17: 40163679-40163679 on Build GRCh38
rs144428076 Chr.17: 40163724-40163724 on Build GRCh38
rs140344787 Chr.17: 40163757-40163757 on Build GRCh38
rs753304836 Chr.17: 40163657-40163657 on Build GRCh38
rs769286027 Chr.17: 40163795-40163795 on Build GRCh38
rs774789844 Chr.17: 40163645-40163645 on Build GRCh38
rs753905501 Chr.17: 40163752-40163752 on Build GRCh38
rs143992032 Chr.17: 40163680-40163680 on Build GRCh38
rs371243053 Chr.17: 40163705-40163705 on Build GRCh38
rs752312436 Chr.17: 40163739-40163739 on Build GRCh38

View Full Product Details