Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 107522176-107522425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALKBH8
Entrez Gene ID
91801
Gene Name
alkB homolog 8, tRNA methyltransferase
Gene Aliases
ABH8, TRM9, TRMT9
UniGene
Hs.503763
Gene Chromosome Location
Chr.11: 107502726-107565740 on Build GRCh38
Transcripts
NM_001301010.1 NM_138775.2 XM_005271728.2 XM_005271729.3 XM_017018553.1 XM_017018554.1 XM_017018555.1 XM_017018556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6894439 c.1261C>G p.L421V Chr.11: 107522325-107522325 on Build GRCh38
4135667 c.1181G>A p.W394* Chr.11: 107522405-107522405 on Build GRCh38
6338886 c.1185G>A p.P395P Chr.11: 107522401-107522401 on Build GRCh38
6894440 c.1261C>G p.L421V Chr.11: 107522325-107522325 on Build GRCh38
4845477 c.1253G>A p.G418E Chr.11: 107522333-107522333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373004685 Chr.11: 107522304-107522304 on Build GRCh38
rs371431984 Chr.11: 107522367-107522367 on Build GRCh38
rs775064272 Chr.11: 107522287-107522287 on Build GRCh38
rs753631187 Chr.11: 107522269-107522269 on Build GRCh38
rs956000961 Chr.11: 107522345-107522345 on Build GRCh38
rs557574244 Chr.11: 107522357-107522357 on Build GRCh38
rs556827511 Chr.11: 107522215-107522215 on Build GRCh38
rs971095710 Chr.11: 107522311-107522311 on Build GRCh38
rs373418821 Chr.11: 107522332-107522332 on Build GRCh38
rs905850520 Chr.11: 107522247-107522247 on Build GRCh38
rs1010781069 Chr.11: 107522395-107522395 on Build GRCh38
rs1006857399 Chr.11: 107522231-107522231 on Build GRCh38
rs907914367 Chr.11: 107522230-107522230 on Build GRCh38
rs765666925 Chr.11: 107522331-107522331 on Build GRCh38
rs568927883 Chr.11: 107522270-107522270 on Build GRCh38
rs747954438 Chr.11: 107522402-107522402 on Build GRCh38
rs754720775 Chr.11: 107522373-107522373 on Build GRCh38
rs768513222 Chr.11: 107522324-107522324 on Build GRCh38
rs1037824508 Chr.11: 107522244-107522244 on Build GRCh38
rs377341673 Chr.11: 107522358-107522358 on Build GRCh38
rs537937283 Chr.11: 107522275-107522275 on Build GRCh38
rs914480436 Chr.11: 107522364-107522364 on Build GRCh38
rs568806408 Chr.11: 107522248-107522248 on Build GRCh38
rs192250667 Chr.11: 107522348-107522348 on Build GRCh38
rs371011851 Chr.11: 107522379-107522379 on Build GRCh38

View Full Product Details