Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 37434771-37435042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf35
Entrez Gene ID
339883
Gene Name
chromosome 3 open reading frame 35
Gene Aliases
APRG1
UniGene
Hs.475945
Gene Chromosome Location
Chr.3: 37379111-37435497 on Build GRCh38
Transcripts
NM_178339.2 NR_126512.1 NR_126513.1 NR_126514.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3945426 c.338G>A p.R113H Chr.3: 37434955-37434955 on Build GRCh38
5452143 c.345C>A p.I115I Chr.3: 37434962-37434962 on Build GRCh38
7310087 c.367A>C p.R123R Chr.3: 37434984-37434984 on Build GRCh38
6227884 c.413C>T p.A138V Chr.3: 37435030-37435030 on Build GRCh38
1044034 c.378C>A p.G126G Chr.3: 37434995-37434995 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769948947 Chr.3: 37434931-37434931 on Build GRCh38
rs375585239 Chr.3: 37434912-37434912 on Build GRCh38
rs749924191 Chr.3: 37434998-37434998 on Build GRCh38
rs757972682 Chr.3: 37435011-37435011 on Build GRCh38
rs758833401 Chr.3: 37434949-37434950 on Build GRCh38
rs779545787 Chr.3: 37435019-37435019 on Build GRCh38
rs371974754 Chr.3: 37434875-37434875 on Build GRCh38
rs759513831 Chr.3: 37434801-37434801 on Build GRCh38
rs973004665 Chr.3: 37434979-37434979 on Build GRCh38
rs756906131 Chr.3: 37434963-37434963 on Build GRCh38
rs753602143 Chr.3: 37434954-37434954 on Build GRCh38
rs756878333 Chr.3: 37434892-37434892 on Build GRCh38
rs777431079 Chr.3: 37434849-37434849 on Build GRCh38
rs374501438 Chr.3: 37434800-37434800 on Build GRCh38
rs368827785 Chr.3: 37434918-37434918 on Build GRCh38
rs202204826 Chr.3: 37435021-37435021 on Build GRCh38
rs577296188 Chr.3: 37434817-37434817 on Build GRCh38
rs13324436 Chr.3: 37434811-37434811 on Build GRCh38
rs201993825 Chr.3: 37434925-37434925 on Build GRCh38
rs780599325 Chr.3: 37434923-37434923 on Build GRCh38
rs767556290 Chr.3: 37434947-37434947 on Build GRCh38
rs555265841 Chr.3: 37435005-37435005 on Build GRCh38
rs372294417 Chr.3: 37434966-37434966 on Build GRCh38
rs776874271 Chr.3: 37434927-37434927 on Build GRCh38
rs200197722 Chr.3: 37434949-37434949 on Build GRCh38
rs760699949 Chr.3: 37434948-37434948 on Build GRCh38
rs17266511 Chr.3: 37434936-37434936 on Build GRCh38
rs192505770 Chr.3: 37434902-37434902 on Build GRCh38
rs757382712 Chr.3: 37434973-37434973 on Build GRCh38
rs143411747 Chr.3: 37434928-37434928 on Build GRCh38
rs200543407 Chr.3: 37434993-37434993 on Build GRCh38
rs559023006 Chr.3: 37434813-37434813 on Build GRCh38
rs961891501 Chr.3: 37434941-37434941 on Build GRCh38
rs896316254 Chr.3: 37434887-37434887 on Build GRCh38
rs183674611 Chr.3: 37434938-37434938 on Build GRCh38
rs746079622 Chr.3: 37434864-37434864 on Build GRCh38
rs544199962 Chr.3: 37434837-37434837 on Build GRCh38
rs954251083 Chr.3: 37434830-37434830 on Build GRCh38
rs747459060 Chr.3: 37435020-37435020 on Build GRCh38

View Full Product Details