Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48771537-48771810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY6
Entrez Gene ID
112
Gene Name
adenylate cyclase 6
Gene Aliases
AC6, LCCS8
UniGene
Hs.525401
Gene Chromosome Location
Chr.12: 48766191-48789096 on Build GRCh38
Transcripts
NM_015270.4 XM_006719210.3 XM_017018743.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6340826 c.2994C>A p.N998K Chr.12: 48771767-48771767 on Build GRCh38
3461416 c.3049G>A p.E1017K Chr.12: 48771712-48771712 on Build GRCh38
2006123 c.3037G>A p.A1013T Chr.12: 48771724-48771724 on Build GRCh38
4649510 c.3027C>T p.N1009N Chr.12: 48771734-48771734 on Build GRCh38
5726705 c.3013C>T p.L1005L Chr.12: 48771748-48771748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143018810 Chr.12: 48771755-48771755 on Build GRCh38
rs759549914 Chr.12: 48771695-48771695 on Build GRCh38
rs761243170 Chr.12: 48771626-48771627 on Build GRCh38
rs370715776 Chr.12: 48771666-48771666 on Build GRCh38
rs1050804189 Chr.12: 48771586-48771586 on Build GRCh38
rs758624938 Chr.12: 48771734-48771734 on Build GRCh38
rs781617475 Chr.12: 48771669-48771669 on Build GRCh38
rs114641886 Chr.12: 48771699-48771699 on Build GRCh38
rs570749264 Chr.12: 48771580-48771580 on Build GRCh38
rs3729972 Chr.12: 48771622-48771622 on Build GRCh38
rs765198318 Chr.12: 48771669-48771670 on Build GRCh38
rs776191450 Chr.12: 48771614-48771615 on Build GRCh38
rs769555800 Chr.12: 48771583-48771583 on Build GRCh38
rs763979879 Chr.12: 48771604-48771604 on Build GRCh38
rs774162371 Chr.12: 48771678-48771678 on Build GRCh38
rs767479214 Chr.12: 48771690-48771690 on Build GRCh38
rs768234224 Chr.12: 48771615-48771616 on Build GRCh38
rs10219743 Chr.12: 48771677-48771677 on Build GRCh38
rs574011369 Chr.12: 48771630-48771630 on Build GRCh38
rs752925722 Chr.12: 48771654-48771654 on Build GRCh38
rs778222597 Chr.12: 48771655-48771655 on Build GRCh38
rs750415828 Chr.12: 48771670-48771670 on Build GRCh38
rs773149602 Chr.12: 48771691-48771691 on Build GRCh38
rs775293381 Chr.12: 48771591-48771591 on Build GRCh38
rs372615937 Chr.12: 48771743-48771743 on Build GRCh38
rs767393565 Chr.12: 48771623-48771623 on Build GRCh38
rs757753382 Chr.12: 48771668-48771668 on Build GRCh38
rs747382194 Chr.12: 48771662-48771662 on Build GRCh38
rs765388900 Chr.12: 48771641-48771641 on Build GRCh38
rs774103755 Chr.12: 48771616-48771616 on Build GRCh38
rs755111509 Chr.12: 48771639-48771639 on Build GRCh38
rs776184259 Chr.12: 48771572-48771572 on Build GRCh38
rs781673810 Chr.12: 48771758-48771758 on Build GRCh38
rs745316597 Chr.12: 48771672-48771672 on Build GRCh38
rs761576657 Chr.12: 48771688-48771688 on Build GRCh38
rs760355693 Chr.12: 48771582-48771590 on Build GRCh38
rs942447979 Chr.12: 48771629-48771629 on Build GRCh38
rs10219585 Chr.12: 48771676-48771676 on Build GRCh38
rs755106681 Chr.12: 48771765-48771765 on Build GRCh38
rs898138570 Chr.12: 48771664-48771664 on Build GRCh38
rs561692050 Chr.12: 48771724-48771724 on Build GRCh38
rs746423786 Chr.12: 48771570-48771570 on Build GRCh38
rs775595042 Chr.12: 48771643-48771643 on Build GRCh38
rs578121356 Chr.12: 48771567-48771567 on Build GRCh38
rs764265386 Chr.12: 48771739-48771739 on Build GRCh38
rs775241937 Chr.12: 48771675-48771675 on Build GRCh38
rs762770111 Chr.12: 48771600-48771600 on Build GRCh38

View Full Product Details