Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44403656-44403905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC5L
Entrez Gene ID
988
Gene Name
cell division cycle 5 like
Gene Aliases
CDC5, CDC5-LIKE, CEF1, PCDC5RP, dJ319D22.1
UniGene
Hs.485471
Gene Chromosome Location
Chr.6: 44387514-44450424 on Build GRCh38
Transcripts
NM_001253.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1582848 c.574C>T p.R192* Chr.6: 44403843-44403843 on Build GRCh38
6624474 c.636C>T p.A212A Chr.6: 44403905-44403905 on Build GRCh38
7196763 c.597G>T p.Q199H Chr.6: 44403866-44403866 on Build GRCh38
5357185 c.540-1G>C p.? Chr.6: 44403808-44403808 on Build GRCh38
3715512 c.611G>A p.R204K Chr.6: 44403880-44403880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761522805 Chr.6: 44403789-44403789 on Build GRCh38
rs921575956 Chr.6: 44403700-44403700 on Build GRCh38
rs751558900 Chr.6: 44403820-44403820 on Build GRCh38
rs200189568 Chr.6: 44403873-44403873 on Build GRCh38
rs773609447 Chr.6: 44403760-44403760 on Build GRCh38
rs766875966 Chr.6: 44403769-44403769 on Build GRCh38
rs1042842148 Chr.6: 44403786-44403786 on Build GRCh38
rs891509190 Chr.6: 44403731-44403731 on Build GRCh38
rs748230177 Chr.6: 44403861-44403861 on Build GRCh38
rs1051459334 Chr.6: 44403730-44403731 on Build GRCh38
rs570813208 Chr.6: 44403777-44403777 on Build GRCh38
rs374359834 Chr.6: 44403790-44403790 on Build GRCh38
rs557219338 Chr.6: 44403706-44403706 on Build GRCh38
rs780093116 Chr.6: 44403798-44403798 on Build GRCh38
rs758159755 Chr.6: 44403796-44403796 on Build GRCh38
rs755044991 Chr.6: 44403831-44403831 on Build GRCh38
rs753421068 Chr.6: 44403778-44403778 on Build GRCh38
rs774868053 Chr.6: 44403774-44403774 on Build GRCh38
rs763273005 Chr.6: 44403761-44403761 on Build GRCh38
rs751926250 Chr.6: 44403843-44403843 on Build GRCh38
rs781421628 Chr.6: 44403842-44403842 on Build GRCh38
rs760128199 Chr.6: 44403776-44403776 on Build GRCh38
rs370695015 Chr.6: 44403877-44403877 on Build GRCh38
rs933120782 Chr.6: 44403713-44403713 on Build GRCh38
rs1010032583 Chr.6: 44403780-44403780 on Build GRCh38

View Full Product Details