Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 122743675-122743908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBS12
Entrez Gene ID
166379
Gene Name
Bardet-Biedl syndrome 12
Gene Aliases
C4orf24
UniGene
Hs.400698
Gene Chromosome Location
Chr.4: 122700437-122744943 on Build GRCh38
Transcripts
NM_001178007.1 NM_152618.2 XM_011531680.2 XM_017007831.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3974707 c.1810T>C p.Y604H Chr.4: 122743702-122743702 on Build GRCh38
4985275 c.1847G>A p.S616N Chr.4: 122743739-122743739 on Build GRCh38
3974708 c.1851A>G p.T617T Chr.4: 122743743-122743743 on Build GRCh38
6207677 c.1923T>C p.S641S Chr.4: 122743815-122743815 on Build GRCh38
243177 c.1998T>C p.V666V Chr.4: 122743890-122743890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753804384 Chr.4: 122743716-122743721 on Build GRCh38
rs748009027 Chr.4: 122743799-122743799 on Build GRCh38
rs765915863 Chr.4: 122743781-122743781 on Build GRCh38
rs112390950 Chr.4: 122743760-122743760 on Build GRCh38
rs778766135 Chr.4: 122743789-122743790 on Build GRCh38
rs369696220 Chr.4: 122743885-122743885 on Build GRCh38
rs773424236 Chr.4: 122743737-122743737 on Build GRCh38
rs141393809 Chr.4: 122743701-122743701 on Build GRCh38
rs939334398 Chr.4: 122743766-122743766 on Build GRCh38
rs28507107 Chr.4: 122743739-122743739 on Build GRCh38
rs781136885 Chr.4: 122743795-122743795 on Build GRCh38
rs184225705 Chr.4: 122743715-122743715 on Build GRCh38
rs528535207 Chr.4: 122743722-122743722 on Build GRCh38
rs778953866 Chr.4: 122743872-122743872 on Build GRCh38
rs772235638 Chr.4: 122743709-122743709 on Build GRCh38
rs139278612 Chr.4: 122743751-122743751 on Build GRCh38
rs147281546 Chr.4: 122743871-122743871 on Build GRCh38
rs771057055 Chr.4: 122743741-122743741 on Build GRCh38
rs779685329 Chr.4: 122743885-122743887 on Build GRCh38
rs756035223 Chr.4: 122743812-122743812 on Build GRCh38
rs758008862 Chr.4: 122743824-122743826 on Build GRCh38
rs892531107 Chr.4: 122743791-122743791 on Build GRCh38
rs759516566 Chr.4: 122743745-122743745 on Build GRCh38
rs777615072 Chr.4: 122743823-122743823 on Build GRCh38
rs757183229 Chr.4: 122743746-122743746 on Build GRCh38
rs745706516 Chr.4: 122743802-122743817 on Build GRCh38
rs758135703 Chr.4: 122743775-122743775 on Build GRCh38
rs938588568 Chr.4: 122743836-122743836 on Build GRCh38
rs751126507 Chr.4: 122743793-122743793 on Build GRCh38
rs931117746 Chr.4: 122743805-122743805 on Build GRCh38
rs764074545 Chr.4: 122743705-122743706 on Build GRCh38
rs749009228 Chr.4: 122743853-122743853 on Build GRCh38
rs779671886 Chr.4: 122743707-122743707 on Build GRCh38
rs376558822 Chr.4: 122743742-122743742 on Build GRCh38
rs764497344 Chr.4: 122743756-122743756 on Build GRCh38
rs17857451 Chr.4: 122743736-122743736 on Build GRCh38
rs13102440 Chr.4: 122743764-122743764 on Build GRCh38
rs776100718 Chr.4: 122743750-122743750 on Build GRCh38
rs905750251 Chr.4: 122743827-122743827 on Build GRCh38
rs369934327 Chr.4: 122743743-122743743 on Build GRCh38
rs768092773 Chr.4: 122743746-122743746 on Build GRCh38

View Full Product Details