Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 68046285-68046510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1D
Entrez Gene ID
10438
Gene Name
C1D nuclear receptor corepressor
Gene Aliases
LRP1, Rrp47, SUN-CoR, SUNCOR, hC1D
UniGene
Hs.602900
Gene Chromosome Location
Chr.2: 68042200-68063027 on Build GRCh38
Transcripts
NM_001190263.1 NM_001190265.1 NM_006333.3 NM_173177.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
419124 c.193T>A p.S65T Chr.2: 68046356-68046356 on Build GRCh38
5653464 c.205G>C p.V69L Chr.2: 68046344-68046344 on Build GRCh38
1022324 c.192T>G p.N64K Chr.2: 68046357-68046357 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767306883 Chr.2: 68046367-68046367 on Build GRCh38
rs779107292 Chr.2: 68046444-68046444 on Build GRCh38
rs913929499 Chr.2: 68046413-68046413 on Build GRCh38
rs750851236 Chr.2: 68046386-68046386 on Build GRCh38
rs752695968 Chr.2: 68046459-68046459 on Build GRCh38
rs745834140 Chr.2: 68046434-68046437 on Build GRCh38
rs746886488 Chr.2: 68046455-68046455 on Build GRCh38
rs567632104 Chr.2: 68046430-68046430 on Build GRCh38
rs775428373 Chr.2: 68046446-68046447 on Build GRCh38
rs751725656 Chr.2: 68046317-68046320 on Build GRCh38
rs756267341 Chr.2: 68046425-68046428 on Build GRCh38
rs767008362 Chr.2: 68046424-68046424 on Build GRCh38
rs779681720 Chr.2: 68046425-68046425 on Build GRCh38
rs140679745 Chr.2: 68046353-68046353 on Build GRCh38
rs936086724 Chr.2: 68046369-68046369 on Build GRCh38
rs748166945 Chr.2: 68046419-68046422 on Build GRCh38
rs1014364076 Chr.2: 68046473-68046473 on Build GRCh38
rs771945369 Chr.2: 68046437-68046440 on Build GRCh38
rs755162995 Chr.2: 68046393-68046393 on Build GRCh38
rs3755168 Chr.2: 68046428-68046428 on Build GRCh38
rs761074195 Chr.2: 68046420-68046420 on Build GRCh38
rs540623850 Chr.2: 68046425-68046426 on Build GRCh38
rs755719262 Chr.2: 68046400-68046400 on Build GRCh38
rs945368085 Chr.2: 68046422-68046433 on Build GRCh38
rs761505425 Chr.2: 68046363-68046363 on Build GRCh38
rs781548875 Chr.2: 68046416-68046416 on Build GRCh38

View Full Product Details