Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 73447160-73447419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDX4
Entrez Gene ID
1046
Gene Name
caudal type homeobox 4
Gene Aliases
-
UniGene
Hs.553488
Gene Chromosome Location
Chr.X: 73447254-73454585 on Build GRCh38
Transcripts
NM_005193.1

SNP Detail

SNP ID Chromosome Location
rs150836467 Chr.X: 73447336-73447336 on Build GRCh38
rs746867826 Chr.X: 73447273-73447273 on Build GRCh38
rs767759680 Chr.X: 73447356-73447356 on Build GRCh38
rs777240076 Chr.X: 73447257-73447257 on Build GRCh38
rs745830531 Chr.X: 73447227-73447228 on Build GRCh38
rs868273091 Chr.X: 73447398-73447398 on Build GRCh38
rs774580628 Chr.X: 73447371-73447371 on Build GRCh38
rs199592731 Chr.X: 73447319-73447319 on Build GRCh38
rs757822096 Chr.X: 73447391-73447391 on Build GRCh38
rs775744896 Chr.X: 73447325-73447325 on Build GRCh38
rs754540125 Chr.X: 73447208-73447208 on Build GRCh38
rs1052035777 Chr.X: 73447192-73447192 on Build GRCh38
rs1007422840 Chr.X: 73447307-73447307 on Build GRCh38
rs886601996 Chr.X: 73447337-73447337 on Build GRCh38
rs758096608 Chr.X: 73447256-73447256 on Build GRCh38
rs375681941 Chr.X: 73447291-73447291 on Build GRCh38
rs773507655 Chr.X: 73447362-73447362 on Build GRCh38
rs768973829 Chr.X: 73447332-73447332 on Build GRCh38
rs756210535 Chr.X: 73447311-73447311 on Build GRCh38
rs770630184 Chr.X: 73447284-73447284 on Build GRCh38
rs778908953 Chr.X: 73447313-73447313 on Build GRCh38
rs778407288 Chr.X: 73447222-73447222 on Build GRCh38
rs367670478 Chr.X: 73447298-73447298 on Build GRCh38
rs761975244 Chr.X: 73447386-73447386 on Build GRCh38
rs532670975 Chr.X: 73447381-73447381 on Build GRCh38
rs761170974 Chr.X: 73447367-73447367 on Build GRCh38
rs752132664 Chr.X: 73447233-73447233 on Build GRCh38
rs940984524 Chr.X: 73447350-73447350 on Build GRCh38
rs370175012 Chr.X: 73447326-73447326 on Build GRCh38
rs768685151 Chr.X: 73447317-73447317 on Build GRCh38

View Full Product Details