Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50892917-50893156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADNP
Entrez Gene ID
23394
Gene Name
activity dependent neuroprotector homeobox
Gene Aliases
ADNP1, HVDAS, MRD28
UniGene
Hs.293736
Gene Chromosome Location
Chr.20: 50888918-50934984 on Build GRCh38
Transcripts
NM_001282531.1 NM_001282532.1 NM_015339.3 NM_181442.2 XM_011528747.2 XM_011528748.2 XM_017027757.1 XM_017027758.1 XM_017027759.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5562699 c.1769C>T p.P590L Chr.20: 50892945-50892945 on Build GRCh38
4793397 c.1754A>G p.N585S Chr.20: 50892960-50892960 on Build GRCh38
1660427 c.1700C>T p.T567I Chr.20: 50893014-50893014 on Build GRCh38
273898 c.1779A>G p.P593P Chr.20: 50892935-50892935 on Build GRCh38
7224190 c.1656G>C p.L552F Chr.20: 50893058-50893058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs909044096 Chr.20: 50893118-50893118 on Build GRCh38
rs776181232 Chr.20: 50893115-50893115 on Build GRCh38
rs369276137 Chr.20: 50893085-50893085 on Build GRCh38
rs760374363 Chr.20: 50892946-50892946 on Build GRCh38
rs756214939 Chr.20: 50893013-50893013 on Build GRCh38
rs920434905 Chr.20: 50893079-50893079 on Build GRCh38
rs921315251 Chr.20: 50893127-50893127 on Build GRCh38
rs371939011 Chr.20: 50893100-50893100 on Build GRCh38
rs147336313 Chr.20: 50892960-50892960 on Build GRCh38
rs138511538 Chr.20: 50892945-50892945 on Build GRCh38
rs376377223 Chr.20: 50893010-50893010 on Build GRCh38
rs146843778 Chr.20: 50893130-50893130 on Build GRCh38
rs769440878 Chr.20: 50893119-50893119 on Build GRCh38
rs139237825 Chr.20: 50892962-50892962 on Build GRCh38
rs867913330 Chr.20: 50892964-50892964 on Build GRCh38
rs751935323 Chr.20: 50892999-50892999 on Build GRCh38
rs746893388 Chr.20: 50893053-50893053 on Build GRCh38
rs376577810 Chr.20: 50893123-50893123 on Build GRCh38
rs144102823 Chr.20: 50892950-50892950 on Build GRCh38
rs745812254 Chr.20: 50893033-50893033 on Build GRCh38
rs746830005 Chr.20: 50893107-50893107 on Build GRCh38
rs943246415 Chr.20: 50893122-50893122 on Build GRCh38
rs573699038 Chr.20: 50893031-50893031 on Build GRCh38
rs772068325 Chr.20: 50893055-50893055 on Build GRCh38
rs369537762 Chr.20: 50893129-50893129 on Build GRCh38
rs761339004 Chr.20: 50893117-50893117 on Build GRCh38
rs968239526 Chr.20: 50892948-50892948 on Build GRCh38
rs1020468395 Chr.20: 50892938-50892938 on Build GRCh38
rs776183933 Chr.20: 50892955-50892955 on Build GRCh38
rs768323837 Chr.20: 50893108-50893108 on Build GRCh38
rs766855083 Chr.20: 50892991-50892991 on Build GRCh38
rs771870178 Chr.20: 50892943-50892943 on Build GRCh38
rs371595931 Chr.20: 50893004-50893004 on Build GRCh38
rs141535413 Chr.20: 50892967-50892967 on Build GRCh38
rs1024149986 Chr.20: 50893024-50893024 on Build GRCh38
rs763350233 Chr.20: 50892977-50892977 on Build GRCh38

View Full Product Details