Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49335391-49335624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD37
Entrez Gene ID
951
Gene Name
CD37 molecule
Gene Aliases
GP52-40, TSPAN26
UniGene
Hs.166556
Gene Chromosome Location
Chr.19: 49335314-49340608 on Build GRCh38
Transcripts
NM_001040031.1 NM_001774.2 XM_005259435.3 XM_005259436.3 XM_011527542.2 XM_011527543.2 XM_011527544.2 XM_017027513.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6003697 c.3G>A p.M1I Chr.19: 49335543-49335543 on Build GRCh38
6003872 c.29T>C p.L10P Chr.19: 49335569-49335569 on Build GRCh38
2752373 c.48C>T p.F16F Chr.19: 49335588-49335588 on Build GRCh38
6003698 c.3G>A p.M1I Chr.19: 49335543-49335543 on Build GRCh38
6003871 c.29T>C p.L10P Chr.19: 49335569-49335569 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533234776 Chr.19: 49335442-49335442 on Build GRCh38
rs767779545 Chr.19: 49335599-49335601 on Build GRCh38
rs200538374 Chr.19: 49335508-49335508 on Build GRCh38
rs982565662 Chr.19: 49335441-49335441 on Build GRCh38
rs369641529 Chr.19: 49335588-49335588 on Build GRCh38
rs769571638 Chr.19: 49335573-49335573 on Build GRCh38
rs757663016 Chr.19: 49335515-49335515 on Build GRCh38
rs992406929 Chr.19: 49335563-49335563 on Build GRCh38
rs887136532 Chr.19: 49335421-49335421 on Build GRCh38
rs764326874 Chr.19: 49335443-49335444 on Build GRCh38
rs200604457 Chr.19: 49335443-49335446 on Build GRCh38
rs773694195 Chr.19: 49335530-49335530 on Build GRCh38
rs561070998 Chr.19: 49335426-49335429 on Build GRCh38
rs373048504 Chr.19: 49335512-49335512 on Build GRCh38
rs767744349 Chr.19: 49335471-49335478 on Build GRCh38
rs530936098 Chr.19: 49335539-49335539 on Build GRCh38
rs979071014 Chr.19: 49335529-49335529 on Build GRCh38
rs908315128 Chr.19: 49335478-49335478 on Build GRCh38
rs371545211 Chr.19: 49335504-49335504 on Build GRCh38
rs1033444393 Chr.19: 49335548-49335548 on Build GRCh38
rs767670119 Chr.19: 49335495-49335495 on Build GRCh38
rs959121468 Chr.19: 49335557-49335557 on Build GRCh38
rs779355702 Chr.19: 49335517-49335517 on Build GRCh38
rs753305889 Chr.19: 49335451-49335452 on Build GRCh38
rs752964901 Chr.19: 49335599-49335599 on Build GRCh38
rs770324194 Chr.19: 49335473-49335473 on Build GRCh38
rs761843933 Chr.19: 49335551-49335551 on Build GRCh38
rs766158685 Chr.19: 49335589-49335589 on Build GRCh38
rs139751919 Chr.19: 49335520-49335520 on Build GRCh38
rs376699678 Chr.19: 49335505-49335505 on Build GRCh38
rs1048096708 Chr.19: 49335415-49335415 on Build GRCh38
rs1038434567 Chr.19: 49335431-49335431 on Build GRCh38
rs771851408 Chr.19: 49335521-49335521 on Build GRCh38
rs752778752 Chr.19: 49335503-49335503 on Build GRCh38
rs143909611 Chr.19: 49335447-49335450 on Build GRCh38
rs768264764 Chr.19: 49335531-49335531 on Build GRCh38
rs11545491 Chr.19: 49335561-49335561 on Build GRCh38

View Full Product Details