Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205523484-205523745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK18
Entrez Gene ID
5129
Gene Name
cyclin dependent kinase 18
Gene Aliases
PCTAIRE, PCTAIRE3, PCTK3
UniGene
Hs.445402
Gene Chromosome Location
Chr.1: 205504556-205532793 on Build GRCh38
Transcripts
NM_002596.3 NM_212502.2 NM_212503.2 XM_011509602.2 XM_017001423.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1661085 c.164C>T p.P55L Chr.1: 205523516-205523516 on Build GRCh38
3789389 c.168G>A p.Q56Q Chr.1: 205523520-205523520 on Build GRCh38
7229631 c.276A>G p.V92V Chr.1: 205523628-205523628 on Build GRCh38
7221171 c.199G>A p.G67R Chr.1: 205523551-205523551 on Build GRCh38
6009750 c.188C>T p.P63L Chr.1: 205523540-205523540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749850106 Chr.1: 205523577-205523577 on Build GRCh38
rs771303595 Chr.1: 205523538-205523538 on Build GRCh38
rs773881781 Chr.1: 205523577-205523577 on Build GRCh38
rs200813019 Chr.1: 205523502-205523502 on Build GRCh38
rs753136425 Chr.1: 205523511-205523511 on Build GRCh38
rs746260003 Chr.1: 205523561-205523561 on Build GRCh38
rs992976696 Chr.1: 205523552-205523552 on Build GRCh38
rs765911809 Chr.1: 205523670-205523670 on Build GRCh38
rs868037490 Chr.1: 205523658-205523658 on Build GRCh38
rs753805512 Chr.1: 205523519-205523519 on Build GRCh38
rs370868675 Chr.1: 205523581-205523581 on Build GRCh38
rs965308424 Chr.1: 205523611-205523611 on Build GRCh38
rs576897417 Chr.1: 205523707-205523707 on Build GRCh38
rs770191177 Chr.1: 205523562-205523562 on Build GRCh38
rs757383800 Chr.1: 205523600-205523600 on Build GRCh38
rs374939365 Chr.1: 205523596-205523596 on Build GRCh38
rs372052332 Chr.1: 205523704-205523704 on Build GRCh38
rs753515361 Chr.1: 205523594-205523594 on Build GRCh38
rs370210105 Chr.1: 205523593-205523593 on Build GRCh38
rs921116057 Chr.1: 205523631-205523631 on Build GRCh38
rs7529442 Chr.1: 205523628-205523628 on Build GRCh38
rs978652420 Chr.1: 205523720-205523720 on Build GRCh38
rs574552377 Chr.1: 205523651-205523651 on Build GRCh38
rs182526430 Chr.1: 205523511-205523511 on Build GRCh38
rs4623769 Chr.1: 205523551-205523551 on Build GRCh38
rs755913151 Chr.1: 205523516-205523516 on Build GRCh38
rs758604272 Chr.1: 205523521-205523521 on Build GRCh38
rs778948701 Chr.1: 205523597-205523597 on Build GRCh38
rs774815207 Chr.1: 205523640-205523640 on Build GRCh38
rs752679057 Chr.1: 205523692-205523692 on Build GRCh38
rs560098318 Chr.1: 205523550-205523550 on Build GRCh38
rs538995199 Chr.1: 205523535-205523535 on Build GRCh38
rs201468269 Chr.1: 205523669-205523669 on Build GRCh38
rs1030180970 Chr.1: 205523666-205523666 on Build GRCh38
rs769171785 Chr.1: 205523636-205523636 on Build GRCh38
rs569695440 Chr.1: 205523517-205523517 on Build GRCh38
rs771603510 Chr.1: 205523650-205523650 on Build GRCh38
rs557040775 Chr.1: 205523677-205523677 on Build GRCh38
rs760450868 Chr.1: 205523586-205523586 on Build GRCh38
rs567773767 Chr.1: 205523644-205523644 on Build GRCh38
rs749612949 Chr.1: 205523633-205523633 on Build GRCh38
rs769083820 Chr.1: 205523569-205523569 on Build GRCh38
rs759385119 Chr.1: 205523674-205523674 on Build GRCh38
rs1020192938 Chr.1: 205523678-205523678 on Build GRCh38
rs867756489 Chr.1: 205523540-205523540 on Build GRCh38
rs35209603 Chr.1: 205523550-205523551 on Build GRCh38
rs377592492 Chr.1: 205523580-205523580 on Build GRCh38
rs781337493 Chr.1: 205523609-205523609 on Build GRCh38
rs949771940 Chr.1: 205523554-205523554 on Build GRCh38
rs368358695 Chr.1: 205523672-205523672 on Build GRCh38
rs778009278 Chr.1: 205523524-205523524 on Build GRCh38
rs769850285 Chr.1: 205523620-205523620 on Build GRCh38
rs990292204 Chr.1: 205523662-205523662 on Build GRCh38
rs946077549 Chr.1: 205523722-205523722 on Build GRCh38
rs368734289 Chr.1: 205523612-205523612 on Build GRCh38
rs763464584 Chr.1: 205523568-205523568 on Build GRCh38
rs756641710 Chr.1: 205523589-205523603 on Build GRCh38
rs765308702 Chr.1: 205523684-205523684 on Build GRCh38

View Full Product Details