Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28766491-28766764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM222B
Entrez Gene ID
55731
Gene Name
family with sequence similarity 222 member B
Gene Aliases
C17orf63
UniGene
Hs.564533
Gene Chromosome Location
Chr.17: 28755978-28855036 on Build GRCh38
Transcripts
NM_001077498.2 NM_001288631.1 NM_001288632.1 NM_001288633.1 NM_001288634.1 NM_001288635.1 NM_001288636.1 NM_001288637.1 NM_001288638.1 NM_001288639.1 NM_001288640.1 NM_018182.3 XM_011524986.2 XM_011524987.1 XM_011524988.2 XM_017024831.1 XM_017024832.1 XM_017024833.1 XM_017024834.1 XM_017024835.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3742146 c.6A>G p.L2L Chr.17: 28766662-28766662 on Build GRCh38
3742147 c.6A>G p.L2L Chr.17: 28766662-28766662 on Build GRCh38
239025 c.55A>G p.T19A Chr.17: 28766613-28766613 on Build GRCh38
6080757 c.55A>T p.T19S Chr.17: 28766613-28766613 on Build GRCh38
6080756 c.55A>T p.T19S Chr.17: 28766613-28766613 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780063525 Chr.17: 28766623-28766623 on Build GRCh38
rs376668197 Chr.17: 28766552-28766552 on Build GRCh38
rs923403264 Chr.17: 28766732-28766732 on Build GRCh38
rs764879442 Chr.17: 28766560-28766560 on Build GRCh38
rs772612037 Chr.17: 28766688-28766688 on Build GRCh38
rs776760246 Chr.17: 28766543-28766543 on Build GRCh38
rs182601873 Chr.17: 28766662-28766662 on Build GRCh38
rs1019559750 Chr.17: 28766637-28766637 on Build GRCh38
rs372131615 Chr.17: 28766718-28766718 on Build GRCh38
rs201191388 Chr.17: 28766590-28766590 on Build GRCh38
rs762898658 Chr.17: 28766573-28766573 on Build GRCh38
rs769592494 Chr.17: 28766709-28766709 on Build GRCh38
rs933773432 Chr.17: 28766734-28766734 on Build GRCh38
rs1029025822 Chr.17: 28766715-28766715 on Build GRCh38
rs756362542 Chr.17: 28766622-28766622 on Build GRCh38
rs9900141 Chr.17: 28766524-28766524 on Build GRCh38
rs370814474 Chr.17: 28766576-28766576 on Build GRCh38
rs748242714 Chr.17: 28766549-28766549 on Build GRCh38
rs908117978 Chr.17: 28766661-28766661 on Build GRCh38
rs568681910 Chr.17: 28766526-28766526 on Build GRCh38
rs751365896 Chr.17: 28766579-28766579 on Build GRCh38
rs775170250 Chr.17: 28766567-28766567 on Build GRCh38
rs536154391 Chr.17: 28766642-28766642 on Build GRCh38
rs367828010 Chr.17: 28766612-28766612 on Build GRCh38
rs757060144 Chr.17: 28766591-28766591 on Build GRCh38
rs368667235 Chr.17: 28766698-28766698 on Build GRCh38
rs780959731 Chr.17: 28766697-28766697 on Build GRCh38
rs748529747 Chr.17: 28766674-28766674 on Build GRCh38
rs770268799 Chr.17: 28766611-28766611 on Build GRCh38
rs770609932 Chr.17: 28766540-28766540 on Build GRCh38
rs965993871 Chr.17: 28766693-28766693 on Build GRCh38
rs755442143 Chr.17: 28766648-28766648 on Build GRCh38

View Full Product Details