Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37808674-37808899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf122
Entrez Gene ID
127687
Gene Name
chromosome 1 open reading frame 122
Gene Aliases
ALAESM
UniGene
Hs.532749
Gene Chromosome Location
Chr.1: 37807801-37809454 on Build GRCh38
Transcripts
NM_001142726.1 NM_198446.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1240922 c.179_181delGAG p.G61delG Chr.1: 37808679-37808681 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs969932092 Chr.1: 37808726-37808726 on Build GRCh38
rs1023994705 Chr.1: 37808706-37808706 on Build GRCh38
rs1038340610 Chr.1: 37808792-37808792 on Build GRCh38
rs907310229 Chr.1: 37808836-37808836 on Build GRCh38
rs781380016 Chr.1: 37808760-37808760 on Build GRCh38
rs760246000 Chr.1: 37808714-37808714 on Build GRCh38
rs778377919 Chr.1: 37808770-37808770 on Build GRCh38
rs764104354 Chr.1: 37808728-37808728 on Build GRCh38
rs547881778 Chr.1: 37808816-37808816 on Build GRCh38
rs757105051 Chr.1: 37808738-37808738 on Build GRCh38
rs191227209 Chr.1: 37808849-37808849 on Build GRCh38
rs538149859 Chr.1: 37808860-37808860 on Build GRCh38
rs773126192 Chr.1: 37808761-37808762 on Build GRCh38
rs761339260 Chr.1: 37808757-37808761 on Build GRCh38
rs750188336 Chr.1: 37808746-37808746 on Build GRCh38
rs925339523 Chr.1: 37808767-37808767 on Build GRCh38
rs529663940 Chr.1: 37808763-37808763 on Build GRCh38
rs756082542 Chr.1: 37808765-37808765 on Build GRCh38
rs749713164 Chr.1: 37808773-37808773 on Build GRCh38
rs764892278 Chr.1: 37808740-37808740 on Build GRCh38
rs948757349 Chr.1: 37808815-37808815 on Build GRCh38
rs954681046 Chr.1: 37808707-37808707 on Build GRCh38
rs919935350 Chr.1: 37808797-37808797 on Build GRCh38
rs758424700 Chr.1: 37808756-37808757 on Build GRCh38
rs753649903 Chr.1: 37808731-37808731 on Build GRCh38
rs765225485 Chr.1: 37808745-37808746 on Build GRCh38
rs979650900 Chr.1: 37808734-37808734 on Build GRCh38
rs755260635 Chr.1: 37808748-37808748 on Build GRCh38
rs780138774 Chr.1: 37808771-37808772 on Build GRCh38
rs906425691 Chr.1: 37808715-37808715 on Build GRCh38

View Full Product Details