Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27137817-27138078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR30
Entrez Gene ID
339779
Gene Name
proline rich 30
Gene Aliases
C2orf53
UniGene
Hs.136555
Gene Chromosome Location
Chr.2: 27136847-27139464 on Build GRCh38
Transcripts
NM_178553.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7358489 c.367C>A p.P123T Chr.2: 27137963-27137963 on Build GRCh38
7358490 c.320G>T p.R107L Chr.2: 27138010-27138010 on Build GRCh38
6091018 c.448G>T p.E150* Chr.2: 27137882-27137882 on Build GRCh38
1407377 c.259T>A p.Y87N Chr.2: 27138071-27138071 on Build GRCh38
3580522 c.332C>T p.S111F Chr.2: 27137998-27137998 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763753142 Chr.2: 27137904-27137904 on Build GRCh38
rs774708771 Chr.2: 27137882-27137882 on Build GRCh38
rs769807431 Chr.2: 27137850-27137850 on Build GRCh38
rs753946403 Chr.2: 27138043-27138043 on Build GRCh38
rs777898103 Chr.2: 27137958-27137958 on Build GRCh38
rs143880024 Chr.2: 27137962-27137962 on Build GRCh38
rs776295484 Chr.2: 27137998-27137998 on Build GRCh38
rs988782459 Chr.2: 27137900-27137900 on Build GRCh38
rs775909372 Chr.2: 27137985-27137985 on Build GRCh38
rs755241813 Chr.2: 27137845-27137845 on Build GRCh38
rs772371217 Chr.2: 27137976-27137976 on Build GRCh38
rs371157472 Chr.2: 27137910-27137910 on Build GRCh38
rs750255025 Chr.2: 27137913-27137913 on Build GRCh38
rs768675313 Chr.2: 27138011-27138011 on Build GRCh38
rs767225905 Chr.2: 27137884-27137884 on Build GRCh38
rs982630511 Chr.2: 27137959-27137960 on Build GRCh38
rs865941721 Chr.2: 27137854-27137854 on Build GRCh38
rs779872335 Chr.2: 27137934-27137934 on Build GRCh38
rs61741796 Chr.2: 27137905-27137905 on Build GRCh38
rs13427842 Chr.2: 27137988-27137988 on Build GRCh38
rs374934991 Chr.2: 27137953-27137953 on Build GRCh38
rs752358736 Chr.2: 27137919-27137921 on Build GRCh38
rs374688258 Chr.2: 27137844-27137844 on Build GRCh38
rs752914740 Chr.2: 27137948-27137948 on Build GRCh38
rs771358963 Chr.2: 27137860-27137860 on Build GRCh38
rs762929202 Chr.2: 27138008-27138008 on Build GRCh38
rs139570211 Chr.2: 27137995-27137995 on Build GRCh38
rs375482932 Chr.2: 27137883-27137883 on Build GRCh38
rs552356273 Chr.2: 27137963-27137963 on Build GRCh38
rs113925617 Chr.2: 27138014-27138014 on Build GRCh38
rs923938001 Chr.2: 27137871-27137871 on Build GRCh38
rs745593024 Chr.2: 27137841-27137841 on Build GRCh38
rs911744446 Chr.2: 27138017-27138017 on Build GRCh38
rs766147706 Chr.2: 27138010-27138010 on Build GRCh38
rs781222541 Chr.2: 27137849-27137849 on Build GRCh38
rs757267683 Chr.2: 27137906-27137906 on Build GRCh38
rs767021340 Chr.2: 27137911-27137912 on Build GRCh38
rs777920249 Chr.2: 27138042-27138042 on Build GRCh38
rs760351915 Chr.2: 27137899-27137899 on Build GRCh38
rs935308761 Chr.2: 27137886-27137886 on Build GRCh38
rs931778766 Chr.2: 27138002-27138002 on Build GRCh38
rs200451312 Chr.2: 27138005-27138005 on Build GRCh38
rs773256177 Chr.2: 27137853-27137853 on Build GRCh38
rs371356602 Chr.2: 27137874-27137874 on Build GRCh38
rs1029144436 Chr.2: 27138024-27138024 on Build GRCh38
rs377737210 Chr.2: 27137895-27137895 on Build GRCh38
rs373438615 Chr.2: 27137842-27137842 on Build GRCh38
rs762858640 Chr.2: 27137972-27137972 on Build GRCh38
rs760357933 Chr.2: 27137927-27137928 on Build GRCh38
rs376483943 Chr.2: 27137857-27137857 on Build GRCh38
rs562865918 Chr.2: 27138004-27138004 on Build GRCh38

View Full Product Details