Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 108884417-108884660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf66
Entrez Gene ID
154907
Gene Name
chromosome 7 open reading frame 66
Gene Aliases
-
UniGene
Hs.276325
Gene Chromosome Location
Chr.7: 108883975-108884587 on Build GRCh38
Transcripts
NM_001024607.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6616661 c.37C>T p.Q13* Chr.7: 108884496-108884496 on Build GRCh38
4475585 c.19C>T p.P7S Chr.7: 108884514-108884514 on Build GRCh38
5799427 c.8C>A p.A3D Chr.7: 108884525-108884525 on Build GRCh38
452150 c.12G>C p.V4V Chr.7: 108884521-108884521 on Build GRCh38
4556382 c.68G>A p.R23K Chr.7: 108884465-108884465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768589012 Chr.7: 108884545-108884545 on Build GRCh38
rs759467504 Chr.7: 108884458-108884458 on Build GRCh38
rs141228190 Chr.7: 108884437-108884437 on Build GRCh38
rs759500066 Chr.7: 108884548-108884548 on Build GRCh38
rs776375398 Chr.7: 108884546-108884546 on Build GRCh38
rs757731554 Chr.7: 108884481-108884481 on Build GRCh38
rs75599531 Chr.7: 108884471-108884471 on Build GRCh38
rs183230116 Chr.7: 108884567-108884567 on Build GRCh38
rs906672949 Chr.7: 108884474-108884474 on Build GRCh38
rs368225324 Chr.7: 108884575-108884575 on Build GRCh38
rs746404116 Chr.7: 108884503-108884503 on Build GRCh38
rs771208852 Chr.7: 108884540-108884540 on Build GRCh38
rs751926747 Chr.7: 108884480-108884480 on Build GRCh38
rs1007658153 Chr.7: 108884516-108884516 on Build GRCh38
rs998079972 Chr.7: 108884580-108884580 on Build GRCh38
rs904033201 Chr.7: 108884515-108884515 on Build GRCh38
rs867907077 Chr.7: 108884587-108884587 on Build GRCh38
rs770262147 Chr.7: 108884504-108884504 on Build GRCh38
rs764374703 Chr.7: 108884576-108884576 on Build GRCh38
rs751839363 Chr.7: 108884579-108884579 on Build GRCh38
rs752773814 Chr.7: 108884466-108884466 on Build GRCh38
rs532788304 Chr.7: 108884441-108884441 on Build GRCh38
rs1029925424 Chr.7: 108884588-108884588 on Build GRCh38
rs760420832 Chr.7: 108884536-108884536 on Build GRCh38
rs142526547 Chr.7: 108884513-108884513 on Build GRCh38
rs893876270 Chr.7: 108884532-108884532 on Build GRCh38
rs760510505 Chr.7: 108884439-108884439 on Build GRCh38
rs1056893898 Chr.7: 108884478-108884478 on Build GRCh38
rs778178126 Chr.7: 108884479-108884479 on Build GRCh38
rs997926171 Chr.7: 108884632-108884632 on Build GRCh38
rs772872504 Chr.7: 108884514-108884514 on Build GRCh38
rs752788360 Chr.7: 108884569-108884569 on Build GRCh38
rs781543768 Chr.7: 108884622-108884622 on Build GRCh38
rs765367894 Chr.7: 108884464-108884464 on Build GRCh38
rs367868708 Chr.7: 108884501-108884501 on Build GRCh38
rs555536391 Chr.7: 108884621-108884621 on Build GRCh38
rs747697932 Chr.7: 108884511-108884511 on Build GRCh38
rs747172103 Chr.7: 108884503-108884504 on Build GRCh38
rs202099108 Chr.7: 108884451-108884451 on Build GRCh38
rs899608595 Chr.7: 108884590-108884590 on Build GRCh38
rs145510967 Chr.7: 108884456-108884456 on Build GRCh38
rs566769968 Chr.7: 108884505-108884505 on Build GRCh38

View Full Product Details