Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69807813-69808086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA4
Entrez Gene ID
307
Gene Name
annexin A4
Gene Aliases
ANX4, HEL-S-274, P32.5, PAP-II, PIG28, PP4-X, ZAP36
UniGene
Hs.422986
Gene Chromosome Location
Chr.2: 69643805-69826477 on Build GRCh38
Transcripts
NM_001153.4 NM_001320698.1 NM_001320700.1 NM_001320702.1 XM_017003943.1 XM_017003944.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6297365 c.397+1G>T p.? Chr.2: 69807997-69807997 on Build GRCh38
5793971 c.372G>T p.R124R Chr.2: 69807971-69807971 on Build GRCh38
1022451 c.353G>A p.R118Q Chr.2: 69807952-69807952 on Build GRCh38
195037 c.374G>A p.R125H Chr.2: 69807973-69807973 on Build GRCh38
4807919 c.397+5G>A p.? Chr.2: 69808001-69808001 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367906194 Chr.2: 69808032-69808032 on Build GRCh38
rs377380903 Chr.2: 69807899-69807899 on Build GRCh38
rs142525768 Chr.2: 69808042-69808042 on Build GRCh38
rs890150558 Chr.2: 69807901-69807901 on Build GRCh38
rs774777486 Chr.2: 69807996-69807996 on Build GRCh38
rs375477161 Chr.2: 69807911-69807911 on Build GRCh38
rs1043015687 Chr.2: 69807846-69807846 on Build GRCh38
rs371456024 Chr.2: 69808010-69808010 on Build GRCh38
rs371185740 Chr.2: 69807910-69807910 on Build GRCh38
rs770362785 Chr.2: 69807973-69807973 on Build GRCh38
rs759205867 Chr.2: 69808001-69808001 on Build GRCh38
rs781597958 Chr.2: 69807968-69807968 on Build GRCh38
rs749388872 Chr.2: 69807989-69807989 on Build GRCh38
rs111370752 Chr.2: 69807882-69807882 on Build GRCh38
rs958078573 Chr.2: 69807935-69807935 on Build GRCh38
rs768786812 Chr.2: 69807991-69807991 on Build GRCh38
rs527727887 Chr.2: 69808030-69808030 on Build GRCh38
rs1033354533 Chr.2: 69807974-69807974 on Build GRCh38
rs757798574 Chr.2: 69808000-69808000 on Build GRCh38
rs757847732 Chr.2: 69807923-69807923 on Build GRCh38
rs1029990697 Chr.2: 69807893-69807893 on Build GRCh38
rs868139393 Chr.2: 69807957-69807957 on Build GRCh38
rs769813390 Chr.2: 69807880-69807880 on Build GRCh38
rs962697042 Chr.2: 69807966-69807966 on Build GRCh38
rs746402744 Chr.2: 69807970-69807970 on Build GRCh38
rs780708282 Chr.2: 69808033-69808033 on Build GRCh38
rs890975561 Chr.2: 69807934-69807934 on Build GRCh38
rs141109795 Chr.2: 69808020-69808020 on Build GRCh38
rs768911507 Chr.2: 69808034-69808034 on Build GRCh38
rs763580819 Chr.2: 69808018-69808018 on Build GRCh38
rs917697989 Chr.2: 69808015-69808015 on Build GRCh38
rs368406744 Chr.2: 69808009-69808009 on Build GRCh38
rs942153342 Chr.2: 69808039-69808039 on Build GRCh38
rs945589513 Chr.2: 69807842-69807842 on Build GRCh38
rs777072688 Chr.2: 69807868-69807868 on Build GRCh38
rs150937606 Chr.2: 69808061-69808061 on Build GRCh38
rs776090398 Chr.2: 69807975-69807975 on Build GRCh38
rs918104078 Chr.2: 69807841-69807841 on Build GRCh38
rs751576696 Chr.2: 69807856-69807856 on Build GRCh38
rs754917686 Chr.2: 69807912-69807912 on Build GRCh38
rs139920396 Chr.2: 69807919-69807919 on Build GRCh38
rs751771934 Chr.2: 69807907-69807907 on Build GRCh38
rs752565943 Chr.2: 69808005-69808005 on Build GRCh38
rs908141953 Chr.2: 69808036-69808036 on Build GRCh38
rs1037821711 Chr.2: 69807849-69807849 on Build GRCh38
rs374028605 Chr.2: 69808021-69808021 on Build GRCh38
rs1015913753 Chr.2: 69807958-69807959 on Build GRCh38
rs900698033 Chr.2: 69808050-69808050 on Build GRCh38
rs765176372 Chr.2: 69808004-69808004 on Build GRCh38
rs144504684 Chr.2: 69807952-69807952 on Build GRCh38
rs774108331 Chr.2: 69808037-69808037 on Build GRCh38
rs746660124 Chr.2: 69807947-69807947 on Build GRCh38
rs778785400 Chr.2: 69807913-69807913 on Build GRCh38
rs760075650 Chr.2: 69807878-69807878 on Build GRCh38
rs374696397 Chr.2: 69807889-69807889 on Build GRCh38
rs777409785 Chr.2: 69807946-69807946 on Build GRCh38
rs757621829 Chr.2: 69807951-69807951 on Build GRCh38
rs762078734 Chr.2: 69807997-69807997 on Build GRCh38
rs745484410 Chr.2: 69807963-69807963 on Build GRCh38
rs766359709 Chr.2: 69807999-69808000 on Build GRCh38

View Full Product Details