Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 26913370-26913631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASUN
Entrez Gene ID
55726
Gene Name
asunder, spermatogenesis regulator
Gene Aliases
C12orf11, GCT1, Mat89Bb, NET48, SPATA30
UniGene
Hs.505077
Gene Chromosome Location
Chr.12: 26905171-26938326 on Build GRCh38
Transcripts
NM_018164.2 XM_005253422.3 XM_011520750.2 XM_011520751.2 XM_011520752.2 XM_011520753.2 XM_017019632.1 XM_017019633.1 XM_017019634.1 XM_017019635.1 XM_017019636.1 XM_017019637.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6606251 c.1682G>A p.R561K Chr.12: 26913580-26913580 on Build GRCh38
6606249 c.1709G>A p.R570Q Chr.12: 26913553-26913553 on Build GRCh38
6326074 c.1701G>A p.E567E Chr.12: 26913561-26913561 on Build GRCh38
215523 c.1717C>T p.R573* Chr.12: 26913545-26913545 on Build GRCh38
938311 c.1637A>G p.N546S Chr.12: 26913625-26913625 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748812805 Chr.12: 26913524-26913524 on Build GRCh38
rs3736227 Chr.12: 26913417-26913417 on Build GRCh38
rs772456262 Chr.12: 26913553-26913553 on Build GRCh38
rs765434863 Chr.12: 26913600-26913600 on Build GRCh38
rs888667810 Chr.12: 26913545-26913545 on Build GRCh38
rs773616300 Chr.12: 26913554-26913554 on Build GRCh38
rs762170385 Chr.12: 26913544-26913544 on Build GRCh38
rs144641134 Chr.12: 26913543-26913543 on Build GRCh38
rs757009397 Chr.12: 26913448-26913448 on Build GRCh38
rs779675707 Chr.12: 26913514-26913514 on Build GRCh38
rs200004956 Chr.12: 26913558-26913558 on Build GRCh38
rs779065936 Chr.12: 26913442-26913442 on Build GRCh38
rs558905071 Chr.12: 26913528-26913528 on Build GRCh38
rs753905740 Chr.12: 26913419-26913419 on Build GRCh38
rs745819814 Chr.12: 26913459-26913459 on Build GRCh38
rs897303948 Chr.12: 26913435-26913435 on Build GRCh38
rs769898393 Chr.12: 26913477-26913477 on Build GRCh38
rs766940470 Chr.12: 26913584-26913584 on Build GRCh38
rs753941687 Chr.12: 26913595-26913595 on Build GRCh38
rs569411628 Chr.12: 26913453-26913453 on Build GRCh38
rs761140099 Chr.12: 26913561-26913561 on Build GRCh38
rs781238391 Chr.12: 26913458-26913458 on Build GRCh38
rs371356263 Chr.12: 26913436-26913436 on Build GRCh38
rs34036164 Chr.12: 26913597-26913597 on Build GRCh38
rs777690730 Chr.12: 26913602-26913602 on Build GRCh38
rs746765518 Chr.12: 26913445-26913445 on Build GRCh38
rs867052401 Chr.12: 26913461-26913461 on Build GRCh38
rs200978962 Chr.12: 26913474-26913474 on Build GRCh38
rs756139464 Chr.12: 26913414-26913414 on Build GRCh38

View Full Product Details