Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13909716-13909989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CC2D1A
Entrez Gene ID
54862
Gene Name
coiled-coil and C2 domain containing 1A
Gene Aliases
FREUD-1, Freud-1/Aki1, MRT3
UniGene
Hs.269592
Gene Chromosome Location
Chr.19: 13906143-13930880 on Build GRCh38
Transcripts
NM_017721.4 XM_005259972.2 XM_005259973.2 XM_005259974.2 XM_005259975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1641063 c.154G>A p.V52I Chr.19: 13909916-13909916 on Build GRCh38
3783186 c.157G>A p.G53R Chr.19: 13909919-13909919 on Build GRCh38
4457005 c.102C>T p.I34I Chr.19: 13909864-13909864 on Build GRCh38
4074715 c.138T>C p.A46A Chr.19: 13909900-13909900 on Build GRCh38
5388768 c.79C>T p.L27F Chr.19: 13909841-13909841 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745363706 Chr.19: 13909739-13909739 on Build GRCh38
rs200254167 Chr.19: 13909918-13909918 on Build GRCh38
rs768454349 Chr.19: 13909785-13909785 on Build GRCh38
rs201404472 Chr.19: 13909843-13909843 on Build GRCh38
rs748987226 Chr.19: 13909783-13909783 on Build GRCh38
rs768366200 Chr.19: 13909873-13909873 on Build GRCh38
rs202235933 Chr.19: 13909930-13909930 on Build GRCh38
rs368933699 Chr.19: 13909910-13909910 on Build GRCh38
rs754032593 Chr.19: 13909816-13909816 on Build GRCh38
rs773270725 Chr.19: 13909801-13909801 on Build GRCh38
rs771963034 Chr.19: 13909883-13909883 on Build GRCh38
rs760634655 Chr.19: 13909899-13909899 on Build GRCh38
rs571264064 Chr.19: 13909942-13909942 on Build GRCh38
rs776630342 Chr.19: 13909898-13909898 on Build GRCh38
rs773968789 Chr.19: 13909791-13909791 on Build GRCh38
rs932225535 Chr.19: 13909761-13909761 on Build GRCh38
rs985559739 Chr.19: 13909766-13909766 on Build GRCh38
rs200450636 Chr.19: 13909821-13909821 on Build GRCh38
rs949890148 Chr.19: 13909838-13909838 on Build GRCh38
rs578166433 Chr.19: 13909920-13909920 on Build GRCh38
rs766322593 Chr.19: 13909897-13909897 on Build GRCh38
rs986294838 Chr.19: 13909770-13909770 on Build GRCh38
rs760733289 Chr.19: 13909802-13909802 on Build GRCh38
rs878936914 Chr.19: 13909933-13909933 on Build GRCh38
rs748349772 Chr.19: 13909749-13909750 on Build GRCh38
rs752002689 Chr.19: 13909919-13909919 on Build GRCh38
rs1001445501 Chr.19: 13909869-13909869 on Build GRCh38
rs774058378 Chr.19: 13909874-13909874 on Build GRCh38
rs369966581 Chr.19: 13909798-13909798 on Build GRCh38
rs769484011 Chr.19: 13909745-13909745 on Build GRCh38
rs12971500 Chr.19: 13909760-13909760 on Build GRCh38
rs752935035 Chr.19: 13909824-13909824 on Build GRCh38
rs761684254 Chr.19: 13909794-13909794 on Build GRCh38
rs755653965 Chr.19: 13909849-13909849 on Build GRCh38
rs748829721 Chr.19: 13909858-13909858 on Build GRCh38
rs966973194 Chr.19: 13909962-13909962 on Build GRCh38
rs779671365 Chr.19: 13909855-13909855 on Build GRCh38
rs766499059 Chr.19: 13909812-13909812 on Build GRCh38
rs375971219 Chr.19: 13909885-13909885 on Build GRCh38
rs758862009 Chr.19: 13909828-13909828 on Build GRCh38
rs878967370 Chr.19: 13909924-13909924 on Build GRCh38
rs910360276 Chr.19: 13909807-13909807 on Build GRCh38
rs773005417 Chr.19: 13909884-13909884 on Build GRCh38
rs747892591 Chr.19: 13909882-13909882 on Build GRCh38
rs765454608 Chr.19: 13909822-13909822 on Build GRCh38
rs772371284 Chr.19: 13909868-13909873 on Build GRCh38
rs775260333 Chr.19: 13909747-13909747 on Build GRCh38
rs765371173 Chr.19: 13909909-13909909 on Build GRCh38
rs1045517051 Chr.19: 13909841-13909841 on Build GRCh38

View Full Product Details