Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 103085128-103085389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDH2
Entrez Gene ID
56898
Gene Name
3-hydroxybutyrate dehydrogenase, type 2
Gene Aliases
DHRS6, EFA6R, PRO20933, SDR15C1, UCPA-OR, UNQ6308
UniGene
Hs.124696
Gene Chromosome Location
Chr.4: 103077618-103099867 on Build GRCh38
Transcripts
NM_020139.3 XM_005263140.3 XM_006714274.3 XM_011532127.2 XM_011532128.2 XM_017008461.1 XM_017008462.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6393822 c.529C>G p.P177A Chr.4: 103085352-103085352 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs528248422 Chr.4: 103085250-103085250 on Build GRCh38
rs772921429 Chr.4: 103085348-103085348 on Build GRCh38
rs763333313 Chr.4: 103085367-103085367 on Build GRCh38
rs534692380 Chr.4: 103085295-103085295 on Build GRCh38
rs765746594 Chr.4: 103085359-103085359 on Build GRCh38
rs548996132 Chr.4: 103085224-103085224 on Build GRCh38
rs760262326 Chr.4: 103085353-103085353 on Build GRCh38
rs201482204 Chr.4: 103085339-103085339 on Build GRCh38
rs761605505 Chr.4: 103085338-103085338 on Build GRCh38
rs768526222 Chr.4: 103085314-103085314 on Build GRCh38
rs922438375 Chr.4: 103085223-103085223 on Build GRCh38
rs7662605 Chr.4: 103085251-103085251 on Build GRCh38
rs975502702 Chr.4: 103085179-103085179 on Build GRCh38
rs764561562 Chr.4: 103085368-103085368 on Build GRCh38
rs7684568 Chr.4: 103085313-103085313 on Build GRCh38
rs913025571 Chr.4: 103085259-103085259 on Build GRCh38
rs749905382 Chr.4: 103085340-103085343 on Build GRCh38
rs1046048367 Chr.4: 103085291-103085291 on Build GRCh38
rs376151940 Chr.4: 103085300-103085300 on Build GRCh38
rs767263970 Chr.4: 103085342-103085342 on Build GRCh38
rs942593752 Chr.4: 103085159-103085159 on Build GRCh38
rs530791412 Chr.4: 103085220-103085220 on Build GRCh38
rs771781292 Chr.4: 103085267-103085267 on Build GRCh38
rs570984130 Chr.4: 103085264-103085264 on Build GRCh38
rs773985286 Chr.4: 103085326-103085326 on Build GRCh38
rs7684344 Chr.4: 103085158-103085158 on Build GRCh38
rs936717091 Chr.4: 103085244-103085244 on Build GRCh38
rs946011147 Chr.4: 103085275-103085275 on Build GRCh38
rs753273018 Chr.4: 103085363-103085363 on Build GRCh38
rs758931763 Chr.4: 103085183-103085183 on Build GRCh38
rs1046827743 Chr.4: 103085230-103085230 on Build GRCh38
rs561180780 Chr.4: 103085227-103085227 on Build GRCh38
rs1039603763 Chr.4: 103085207-103085207 on Build GRCh38

View Full Product Details