Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201208022-201208277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP10
Entrez Gene ID
843
Gene Name
caspase 10
Gene Aliases
ALPS2, FLICE2, MCH4
UniGene
Hs.5353
Gene Chromosome Location
Chr.2: 201182881-201229406 on Build GRCh38
Transcripts
NM_001206524.1 NM_001206542.1 NM_001230.4 NM_032974.4 NM_032976.3 NM_032977.3 XM_005246907.2 XM_006712796.3 XM_011511990.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5861487 c.874C>T p.H292Y Chr.2: 201208135-201208135 on Build GRCh38
3909328 c.852C>T p.G284G Chr.2: 201208113-201208113 on Build GRCh38
4764375 c.922+1G>A p.? Chr.2: 201208184-201208184 on Build GRCh38
3909329 c.852C>T p.G284G Chr.2: 201208113-201208113 on Build GRCh38
4764376 c.922+1G>A p.? Chr.2: 201208184-201208184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367848313 Chr.2: 201208221-201208221 on Build GRCh38
rs762814898 Chr.2: 201208059-201208059 on Build GRCh38
rs769651296 Chr.2: 201208177-201208178 on Build GRCh38
rs146654699 Chr.2: 201208148-201208148 on Build GRCh38
rs559244194 Chr.2: 201208188-201208188 on Build GRCh38
rs768371369 Chr.2: 201208170-201208170 on Build GRCh38
rs373189673 Chr.2: 201208206-201208206 on Build GRCh38
rs746636209 Chr.2: 201208124-201208124 on Build GRCh38
rs981465275 Chr.2: 201208172-201208172 on Build GRCh38
rs113096171 Chr.2: 201208211-201208211 on Build GRCh38
rs748988477 Chr.2: 201208171-201208171 on Build GRCh38
rs756914853 Chr.2: 201208218-201208218 on Build GRCh38
rs753175958 Chr.2: 201208215-201208215 on Build GRCh38
rs762072082 Chr.2: 201208069-201208069 on Build GRCh38
rs936253557 Chr.2: 201208162-201208162 on Build GRCh38
rs113867411 Chr.2: 201208077-201208077 on Build GRCh38
rs760814428 Chr.2: 201208200-201208200 on Build GRCh38
rs958667368 Chr.2: 201208091-201208091 on Build GRCh38
rs775531682 Chr.2: 201208155-201208155 on Build GRCh38
rs757006791 Chr.2: 201208128-201208128 on Build GRCh38
rs774579828 Chr.2: 201208065-201208065 on Build GRCh38
rs769831223 Chr.2: 201208053-201208053 on Build GRCh38
rs764264041 Chr.2: 201208060-201208060 on Build GRCh38
rs111719534 Chr.2: 201208117-201208117 on Build GRCh38
rs372906837 Chr.2: 201208107-201208107 on Build GRCh38
rs200221261 Chr.2: 201208184-201208184 on Build GRCh38
rs774173689 Chr.2: 201208177-201208177 on Build GRCh38
rs745398864 Chr.2: 201208141-201208141 on Build GRCh38
rs540207844 Chr.2: 201208158-201208158 on Build GRCh38
rs767555790 Chr.2: 201208075-201208075 on Build GRCh38
rs149912574 Chr.2: 201208174-201208174 on Build GRCh38
rs750581097 Chr.2: 201208100-201208100 on Build GRCh38
rs369897442 Chr.2: 201208140-201208140 on Build GRCh38
rs991257476 Chr.2: 201208106-201208106 on Build GRCh38
rs753572822 Chr.2: 201208209-201208209 on Build GRCh38
rs17860403 Chr.2: 201208114-201208114 on Build GRCh38
rs775092096 Chr.2: 201208055-201208055 on Build GRCh38
rs773105887 Chr.2: 201208194-201208194 on Build GRCh38

View Full Product Details