Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 88045519-88045792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf29
Entrez Gene ID
91298
Gene Name
chromosome 12 open reading frame 29
Gene Aliases
-
UniGene
Hs.591009
Gene Chromosome Location
Chr.12: 88035491-88050160 on Build GRCh38
Transcripts
NM_001009894.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6614162 c.498C>T p.A166A Chr.12: 88045658-88045658 on Build GRCh38
4045253 c.463C>T p.H155Y Chr.12: 88045623-88045623 on Build GRCh38
1188562 c.551T>G p.V184G Chr.12: 88045711-88045711 on Build GRCh38
5920358 c.427G>A p.V143I Chr.12: 88045587-88045587 on Build GRCh38
116433 c.496G>A p.A166T Chr.12: 88045656-88045656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764242842 Chr.12: 88045613-88045613 on Build GRCh38
rs746163866 Chr.12: 88045695-88045695 on Build GRCh38
rs982025816 Chr.12: 88045750-88045750 on Build GRCh38
rs766195578 Chr.12: 88045627-88045627 on Build GRCh38
rs765324047 Chr.12: 88045765-88045765 on Build GRCh38
rs772734581 Chr.12: 88045703-88045703 on Build GRCh38
rs200707946 Chr.12: 88045549-88045549 on Build GRCh38
rs775712497 Chr.12: 88045587-88045587 on Build GRCh38
rs374005744 Chr.12: 88045544-88045544 on Build GRCh38
rs146104771 Chr.12: 88045723-88045723 on Build GRCh38
rs774791106 Chr.12: 88045690-88045690 on Build GRCh38
rs1030382038 Chr.12: 88045599-88045599 on Build GRCh38
rs777328255 Chr.12: 88045667-88045667 on Build GRCh38
rs368654905 Chr.12: 88045545-88045545 on Build GRCh38
rs760793897 Chr.12: 88045608-88045608 on Build GRCh38
rs748159531 Chr.12: 88045556-88045559 on Build GRCh38
rs760106770 Chr.12: 88045582-88045582 on Build GRCh38
rs754071865 Chr.12: 88045619-88045619 on Build GRCh38
rs762085199 Chr.12: 88045755-88045755 on Build GRCh38
rs751559637 Chr.12: 88045633-88045633 on Build GRCh38
rs994008316 Chr.12: 88045560-88045560 on Build GRCh38
rs769616634 Chr.12: 88045671-88045673 on Build GRCh38
rs536756040 Chr.12: 88045640-88045640 on Build GRCh38
rs773118358 Chr.12: 88045705-88045705 on Build GRCh38
rs368001947 Chr.12: 88045578-88045578 on Build GRCh38
rs760883831 Chr.12: 88045727-88045727 on Build GRCh38
rs771294569 Chr.12: 88045559-88045559 on Build GRCh38
rs768079805 Chr.12: 88045585-88045585 on Build GRCh38
rs371802467 Chr.12: 88045622-88045622 on Build GRCh38
rs181387601 Chr.12: 88045551-88045551 on Build GRCh38
rs748964838 Chr.12: 88045676-88045676 on Build GRCh38
rs369007823 Chr.12: 88045646-88045646 on Build GRCh38
rs771257359 Chr.12: 88045686-88045686 on Build GRCh38
rs755603808 Chr.12: 88045662-88045662 on Build GRCh38
rs1017598300 Chr.12: 88045739-88045739 on Build GRCh38
rs375787531 Chr.12: 88045644-88045644 on Build GRCh38
rs764205685 Chr.12: 88045730-88045730 on Build GRCh38
rs776788090 Chr.12: 88045731-88045731 on Build GRCh38

View Full Product Details