Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 10262410-10262683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT5
Entrez Gene ID
22948
Gene Name
chaperonin containing TCP1 subunit 5
Gene Aliases
CCT-epsilon, CCTE, HEL-S-69, PNAS-102, TCP-1-epsilon
UniGene
Hs.1600
Gene Chromosome Location
Chr.5: 10249921-10266412 on Build GRCh38
Transcripts
NM_001306153.1 NM_001306154.1 NM_001306155.1 NM_001306156.1 NM_012073.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623311 c.1240C>T p.R414C Chr.5: 10262541-10262541 on Build GRCh38
4749836 c.1277A>G p.E426G Chr.5: 10262578-10262578 on Build GRCh38
3409651 c.1240C>A p.R414S Chr.5: 10262541-10262541 on Build GRCh38
5455337 c.1193C>T p.A398V Chr.5: 10262494-10262494 on Build GRCh38
381024 c.1230G>T p.R410R Chr.5: 10262531-10262531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767982457 Chr.5: 10262571-10262571 on Build GRCh38
rs768166472 Chr.5: 10262462-10262462 on Build GRCh38
rs923206809 Chr.5: 10262529-10262529 on Build GRCh38
rs771944850 Chr.5: 10262530-10262530 on Build GRCh38
rs199800854 Chr.5: 10262555-10262555 on Build GRCh38
rs761273276 Chr.5: 10262551-10262551 on Build GRCh38
rs774663416 Chr.5: 10262511-10262511 on Build GRCh38
rs757459371 Chr.5: 10262590-10262590 on Build GRCh38
rs748625875 Chr.5: 10262560-10262560 on Build GRCh38
rs746171086 Chr.5: 10262598-10262598 on Build GRCh38
rs576837156 Chr.5: 10262591-10262591 on Build GRCh38
rs1024096578 Chr.5: 10262620-10262620 on Build GRCh38
rs1000196897 Chr.5: 10262640-10262640 on Build GRCh38
rs771204776 Chr.5: 10262471-10262471 on Build GRCh38
rs763731217 Chr.5: 10262440-10262440 on Build GRCh38
rs11557654 Chr.5: 10262544-10262544 on Build GRCh38
rs80133586 Chr.5: 10262444-10262444 on Build GRCh38
rs1020211399 Chr.5: 10262621-10262621 on Build GRCh38
rs149067488 Chr.5: 10262611-10262611 on Build GRCh38
rs772175136 Chr.5: 10262609-10262609 on Build GRCh38
rs756917554 Chr.5: 10262461-10262461 on Build GRCh38
rs545733096 Chr.5: 10262626-10262626 on Build GRCh38
rs201280643 Chr.5: 10262584-10262584 on Build GRCh38
rs773613720 Chr.5: 10262651-10262651 on Build GRCh38
rs2578642 Chr.5: 10262628-10262628 on Build GRCh38
rs201810364 Chr.5: 10262494-10262494 on Build GRCh38
rs746067421 Chr.5: 10262517-10262517 on Build GRCh38
rs768671276 Chr.5: 10262617-10262617 on Build GRCh38
rs762679462 Chr.5: 10262652-10262652 on Build GRCh38
rs201013299 Chr.5: 10262538-10262538 on Build GRCh38
rs781378409 Chr.5: 10262484-10262484 on Build GRCh38
rs750114723 Chr.5: 10262558-10262558 on Build GRCh38
rs369196153 Chr.5: 10262596-10262596 on Build GRCh38
rs766680212 Chr.5: 10262659-10262659 on Build GRCh38
rs753337234 Chr.5: 10262447-10262447 on Build GRCh38
rs771307325 Chr.5: 10262510-10262510 on Build GRCh38
rs748756363 Chr.5: 10262492-10262492 on Build GRCh38
rs372827828 Chr.5: 10262469-10262469 on Build GRCh38
rs187490382 Chr.5: 10262539-10262539 on Build GRCh38
rs375481646 Chr.5: 10262632-10262632 on Build GRCh38
rs752947134 Chr.5: 10262573-10262573 on Build GRCh38
rs968539902 Chr.5: 10262449-10262449 on Build GRCh38
rs776123781 Chr.5: 10262550-10262550 on Build GRCh38
rs760501927 Chr.5: 10262438-10262438 on Build GRCh38
rs755417554 Chr.5: 10262483-10262483 on Build GRCh38
rs10474867 Chr.5: 10262658-10262658 on Build GRCh38
rs756645693 Chr.5: 10262575-10262575 on Build GRCh38
rs751954199 Chr.5: 10262477-10262477 on Build GRCh38
rs144940167 Chr.5: 10262546-10262546 on Build GRCh38
rs746715722 Chr.5: 10262612-10262612 on Build GRCh38
rs1139515 Chr.5: 10262595-10262595 on Build GRCh38
rs113262482 Chr.5: 10262495-10262495 on Build GRCh38
rs943286316 Chr.5: 10262507-10262507 on Build GRCh38
rs749310473 Chr.5: 10262500-10262500 on Build GRCh38
rs964509347 Chr.5: 10262660-10262660 on Build GRCh38
rs200430696 Chr.5: 10262541-10262541 on Build GRCh38

View Full Product Details