Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 98665638-98665875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APAF1
Entrez Gene ID
317
Gene Name
apoptotic peptidase activating factor 1
Gene Aliases
APAF-1, CED4
UniGene
Hs.552567
Gene Chromosome Location
Chr.12: 98645141-98735433 on Build GRCh38
Transcripts
NM_001160.2 NM_013229.2 NM_181861.1 NM_181868.1 NM_181869.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3813294 c.1086G>T p.E362D Chr.12: 98665683-98665683 on Build GRCh38
5493009 c.1176C>T p.D392D Chr.12: 98665773-98665773 on Build GRCh38
944891 c.1055G>T p.R352I Chr.12: 98665652-98665652 on Build GRCh38
7173999 c.1078G>A p.D360N Chr.12: 98665675-98665675 on Build GRCh38
6478468 c.1063A>G p.K355E Chr.12: 98665660-98665660 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141824871 Chr.12: 98665723-98665723 on Build GRCh38
rs770350894 Chr.12: 98665724-98665724 on Build GRCh38
rs773700554 Chr.12: 98665726-98665726 on Build GRCh38
rs748600489 Chr.12: 98665718-98665718 on Build GRCh38
rs757171141 Chr.12: 98665788-98665788 on Build GRCh38
rs57171247 Chr.12: 98665674-98665674 on Build GRCh38
rs768162508 Chr.12: 98665705-98665705 on Build GRCh38
rs774596730 Chr.12: 98665761-98665761 on Build GRCh38
rs754450904 Chr.12: 98665816-98665816 on Build GRCh38
rs200279687 Chr.12: 98665821-98665821 on Build GRCh38
rs758148327 Chr.12: 98665808-98665808 on Build GRCh38
rs980221719 Chr.12: 98665680-98665680 on Build GRCh38
rs767562909 Chr.12: 98665785-98665785 on Build GRCh38
rs778671229 Chr.12: 98665671-98665671 on Build GRCh38
rs201629052 Chr.12: 98665730-98665730 on Build GRCh38
rs913525980 Chr.12: 98665801-98665801 on Build GRCh38
rs753782480 Chr.12: 98665786-98665786 on Build GRCh38
rs766680485 Chr.12: 98665747-98665747 on Build GRCh38
rs779594536 Chr.12: 98665809-98665809 on Build GRCh38
rs376065572 Chr.12: 98665807-98665807 on Build GRCh38
rs927344084 Chr.12: 98665742-98665742 on Build GRCh38
rs777017529 Chr.12: 98665710-98665710 on Build GRCh38
rs746614623 Chr.12: 98665812-98665812 on Build GRCh38
rs201932795 Chr.12: 98665692-98665692 on Build GRCh38
rs765064029 Chr.12: 98665804-98665804 on Build GRCh38
rs570250043 Chr.12: 98665774-98665774 on Build GRCh38
rs758021604 Chr.12: 98665676-98665676 on Build GRCh38
rs768708171 Chr.12: 98665766-98665768 on Build GRCh38
rs780750349 Chr.12: 98665817-98665817 on Build GRCh38
rs779653660 Chr.12: 98665678-98665678 on Build GRCh38
rs1050185397 Chr.12: 98665839-98665839 on Build GRCh38
rs746490911 Chr.12: 98665694-98665694 on Build GRCh38
rs201107103 Chr.12: 98665818-98665818 on Build GRCh38

View Full Product Details