Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 24978794-24979019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APMAP
Entrez Gene ID
57136
Gene Name
adipocyte plasma membrane associated protein
Gene Aliases
BSCv, C20orf3
UniGene
Hs.472330
Gene Chromosome Location
Chr.20: 24962925-24992974 on Build GRCh38
Transcripts
NM_020531.2 XM_005260763.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1025301 c.274G>C p.E92Q Chr.20: 24978821-24978821 on Build GRCh38
6092977 c.244C>G p.L82V Chr.20: 24978851-24978851 on Build GRCh38
4819875 c.274G>A p.E92K Chr.20: 24978821-24978821 on Build GRCh38
6602242 c.223C>T p.P75S Chr.20: 24978872-24978872 on Build GRCh38
6414141 c.265C>T p.R89* Chr.20: 24978830-24978830 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936935503 Chr.20: 24978915-24978915 on Build GRCh38
rs151079082 Chr.20: 24978842-24978842 on Build GRCh38
rs189459322 Chr.20: 24978973-24978973 on Build GRCh38
rs527748001 Chr.20: 24978867-24978867 on Build GRCh38
rs1054468409 Chr.20: 24978947-24978947 on Build GRCh38
rs777419745 Chr.20: 24978922-24978922 on Build GRCh38
rs771498788 Chr.20: 24978913-24978913 on Build GRCh38
rs373359836 Chr.20: 24978908-24978908 on Build GRCh38
rs761936268 Chr.20: 24978847-24978847 on Build GRCh38
rs759889840 Chr.20: 24978830-24978830 on Build GRCh38
rs202015003 Chr.20: 24978937-24978937 on Build GRCh38
rs577201722 Chr.20: 24978839-24978839 on Build GRCh38
rs200857426 Chr.20: 24978855-24978855 on Build GRCh38
rs980052521 Chr.20: 24978975-24978975 on Build GRCh38
rs775937714 Chr.20: 24978831-24978831 on Build GRCh38
rs755010382 Chr.20: 24978896-24978896 on Build GRCh38
rs780023247 Chr.20: 24978882-24978882 on Build GRCh38
rs141100068 Chr.20: 24978868-24978868 on Build GRCh38
rs771828282 Chr.20: 24978823-24978823 on Build GRCh38
rs917331961 Chr.20: 24978945-24978945 on Build GRCh38
rs753776971 Chr.20: 24978894-24978894 on Build GRCh38
rs961485029 Chr.20: 24978914-24978914 on Build GRCh38
rs754643037 Chr.20: 24978979-24978979 on Build GRCh38
rs746696923 Chr.20: 24978926-24978926 on Build GRCh38
rs556885938 Chr.20: 24978983-24978983 on Build GRCh38
rs778287087 Chr.20: 24978898-24978898 on Build GRCh38
rs776849853 Chr.20: 24978829-24978829 on Build GRCh38
rs565873771 Chr.20: 24978862-24978862 on Build GRCh38
rs754994056 Chr.20: 24978838-24978838 on Build GRCh38

View Full Product Details