Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143299850-143300083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP2
Entrez Gene ID
835
Gene Name
caspase 2
Gene Aliases
CASP-2, ICH1, NEDD-2, NEDD2, PPP1R57
UniGene
Hs.368982
Gene Chromosome Location
Chr.7: 143288215-143307696 on Build GRCh38
Transcripts
NM_001224.4 NM_032982.3 NM_032983.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1086730 c.843C>T p.G281G Chr.7: 143300018-143300018 on Build GRCh38
4153924 c.871C>T p.L291F Chr.7: 143300046-143300046 on Build GRCh38
1086729 c.828delG p.H277fs*80 Chr.7: 143300003-143300003 on Build GRCh38
1086728 c.777A>G p.A259A Chr.7: 143299952-143299952 on Build GRCh38
1635166 c.811A>G p.I271V Chr.7: 143299986-143299986 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747906301 Chr.7: 143299983-143299983 on Build GRCh38
rs746604671 Chr.7: 143299995-143299995 on Build GRCh38
rs751755859 Chr.7: 143299940-143299940 on Build GRCh38
rs370387103 Chr.7: 143300056-143300056 on Build GRCh38
rs763405941 Chr.7: 143300019-143300019 on Build GRCh38
rs376495263 Chr.7: 143299896-143299896 on Build GRCh38
rs145310616 Chr.7: 143300060-143300060 on Build GRCh38
rs951610625 Chr.7: 143299934-143299934 on Build GRCh38
rs754893292 Chr.7: 143299975-143299975 on Build GRCh38
rs766926410 Chr.7: 143299893-143299893 on Build GRCh38
rs760024653 Chr.7: 143299899-143299899 on Build GRCh38
rs114290247 Chr.7: 143299976-143299976 on Build GRCh38
rs754311596 Chr.7: 143299894-143299894 on Build GRCh38
rs759986667 Chr.7: 143300005-143300005 on Build GRCh38
rs200779188 Chr.7: 143299988-143299988 on Build GRCh38
rs765756378 Chr.7: 143300009-143300009 on Build GRCh38
rs763167453 Chr.7: 143300011-143300011 on Build GRCh38
rs898132028 Chr.7: 143299892-143299892 on Build GRCh38
rs1019753057 Chr.7: 143299943-143299943 on Build GRCh38
rs539015555 Chr.7: 143300046-143300046 on Build GRCh38
rs1055400752 Chr.7: 143299885-143299885 on Build GRCh38
rs757402027 Chr.7: 143299967-143299967 on Build GRCh38
rs150852779 Chr.7: 143299968-143299968 on Build GRCh38
rs143669356 Chr.7: 143299937-143299937 on Build GRCh38
rs764470760 Chr.7: 143300027-143300027 on Build GRCh38
rs370097426 Chr.7: 143299989-143299989 on Build GRCh38
rs758747020 Chr.7: 143299932-143299932 on Build GRCh38
rs367713957 Chr.7: 143300018-143300018 on Build GRCh38
rs776908061 Chr.7: 143299951-143299952 on Build GRCh38
rs373653048 Chr.7: 143300003-143300003 on Build GRCh38
rs139233385 Chr.7: 143299969-143299969 on Build GRCh38
rs753060444 Chr.7: 143299931-143299931 on Build GRCh38
rs777042580 Chr.7: 143299911-143299911 on Build GRCh38

View Full Product Details