Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231225366-231225623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf131
Entrez Gene ID
128061
Gene Name
chromosome 1 open reading frame 131
Gene Aliases
-
UniGene
Hs.743464
Gene Chromosome Location
Chr.1: 231223763-231241187 on Build GRCh38
Transcripts
NM_001300830.1 NM_152379.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3770249 c.741_743delGGA p.E247delE Chr.1: 231225474-231225476 on Build GRCh38
1340068 c.746G>T p.R249M Chr.1: 231225471-231225471 on Build GRCh38
6323512 c.741G>A p.E247E Chr.1: 231225476-231225476 on Build GRCh38
6615088 c.700G>T p.E234* Chr.1: 231225517-231225517 on Build GRCh38
6287677 c.707A>C p.D236A Chr.1: 231225510-231225510 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756719616 Chr.1: 231225542-231225542 on Build GRCh38
rs780033208 Chr.1: 231225551-231225551 on Build GRCh38
rs778623026 Chr.1: 231225418-231225418 on Build GRCh38
rs768603926 Chr.1: 231225452-231225452 on Build GRCh38
rs763322144 Chr.1: 231225505-231225505 on Build GRCh38
rs369953866 Chr.1: 231225420-231225420 on Build GRCh38
rs552622716 Chr.1: 231225518-231225518 on Build GRCh38
rs1013471540 Chr.1: 231225581-231225581 on Build GRCh38
rs367764624 Chr.1: 231225402-231225402 on Build GRCh38
rs766681039 Chr.1: 231225507-231225507 on Build GRCh38
rs202069249 Chr.1: 231225558-231225558 on Build GRCh38
rs1043187477 Chr.1: 231225508-231225508 on Build GRCh38
rs372471370 Chr.1: 231225435-231225435 on Build GRCh38
rs746775512 Chr.1: 231225565-231225565 on Build GRCh38
rs758295886 Chr.1: 231225550-231225550 on Build GRCh38
rs1037896585 Chr.1: 231225450-231225450 on Build GRCh38
rs769640246 Chr.1: 231225495-231225495 on Build GRCh38
rs775585839 Chr.1: 231225442-231225442 on Build GRCh38
rs776626538 Chr.1: 231225454-231225454 on Build GRCh38
rs750595253 Chr.1: 231225490-231225490 on Build GRCh38
rs768536974 Chr.1: 231225571-231225571 on Build GRCh38
rs377041638 Chr.1: 231225478-231225478 on Build GRCh38
rs753354101 Chr.1: 231225523-231225523 on Build GRCh38
rs765487858 Chr.1: 231225501-231225501 on Build GRCh38
rs778442739 Chr.1: 231225546-231225546 on Build GRCh38
rs761941983 Chr.1: 231225469-231225469 on Build GRCh38
rs969416291 Chr.1: 231225389-231225389 on Build GRCh38
rs780451045 Chr.1: 231225474-231225476 on Build GRCh38
rs747901029 Chr.1: 231225486-231225491 on Build GRCh38
rs752093961 Chr.1: 231225514-231225514 on Build GRCh38
rs912841039 Chr.1: 231225404-231225404 on Build GRCh38
rs570122590 Chr.1: 231225392-231225392 on Build GRCh38
rs761238949 Chr.1: 231225474-231225474 on Build GRCh38
rs1022421616 Chr.1: 231225591-231225591 on Build GRCh38
rs369195842 Chr.1: 231225548-231225548 on Build GRCh38
rs947413917 Chr.1: 231225500-231225500 on Build GRCh38
rs755521484 Chr.1: 231225516-231225516 on Build GRCh38
rs940456343 Chr.1: 231225444-231225444 on Build GRCh38
rs896386444 Chr.1: 231225557-231225557 on Build GRCh38
rs760646719 Chr.1: 231225446-231225446 on Build GRCh38
rs903175951 Chr.1: 231225517-231225517 on Build GRCh38

View Full Product Details