Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 43492250-43492475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf34
Entrez Gene ID
375444
Gene Name
chromosome 5 open reading frame 34
Gene Aliases
-
UniGene
Hs.661641
Gene Chromosome Location
Chr.5: 43486699-43515433 on Build GRCh38
Transcripts
NM_198566.3 XM_011514038.2 XM_017009445.1 XM_017009446.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6350054 c.1531G>A p.D511N Chr.5: 43492264-43492264 on Build GRCh38
1620343 c.1495G>C p.G499R Chr.5: 43492300-43492300 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749919469 Chr.5: 43492323-43492323 on Build GRCh38
rs1000197387 Chr.5: 43492425-43492425 on Build GRCh38
rs888062432 Chr.5: 43492370-43492370 on Build GRCh38
rs773299218 Chr.5: 43492291-43492291 on Build GRCh38
rs1034429108 Chr.5: 43492437-43492437 on Build GRCh38
rs755774169 Chr.5: 43492324-43492324 on Build GRCh38
rs780223026 Chr.5: 43492289-43492293 on Build GRCh38
rs753217928 Chr.5: 43492317-43492317 on Build GRCh38
rs1007819780 Chr.5: 43492432-43492432 on Build GRCh38
rs772296889 Chr.5: 43492277-43492277 on Build GRCh38
rs373958505 Chr.5: 43492325-43492325 on Build GRCh38
rs369516105 Chr.5: 43492311-43492311 on Build GRCh38
rs778797452 Chr.5: 43492352-43492352 on Build GRCh38
rs1049058593 Chr.5: 43492366-43492366 on Build GRCh38
rs768637972 Chr.5: 43492322-43492322 on Build GRCh38
rs1020186298 Chr.5: 43492357-43492357 on Build GRCh38
rs138845538 Chr.5: 43492294-43492294 on Build GRCh38
rs748122653 Chr.5: 43492356-43492356 on Build GRCh38
rs748034742 Chr.5: 43492275-43492275 on Build GRCh38
rs766028877 Chr.5: 43492444-43492444 on Build GRCh38
rs1045866149 Chr.5: 43492303-43492303 on Build GRCh38
rs765518351 Chr.5: 43492316-43492316 on Build GRCh38
rs767046979 Chr.5: 43492322-43492322 on Build GRCh38
rs776917143 Chr.5: 43492306-43492306 on Build GRCh38
rs771159760 Chr.5: 43492299-43492299 on Build GRCh38
rs747036823 Chr.5: 43492299-43492299 on Build GRCh38
rs754829101 Chr.5: 43492339-43492339 on Build GRCh38
rs753646027 Chr.5: 43492328-43492328 on Build GRCh38
rs904544070 Chr.5: 43492363-43492366 on Build GRCh38

View Full Product Details