Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51291905-51292176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIN2
Entrez Gene ID
51411
Gene Name
bridging integrator 2
Gene Aliases
BRAP-1
UniGene
Hs.14770
Gene Chromosome Location
Chr.12: 51281038-51324680 on Build GRCh38
Transcripts
NM_001290007.1 NM_001290008.1 NM_001290009.1 NM_016293.3 XM_005268957.3 XM_005268958.3 XM_017019422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7003131 c.1145C>G p.S382* Chr.12: 51291961-51291961 on Build GRCh38
7272629 c.1047C>T p.S349S Chr.12: 51291981-51291981 on Build GRCh38
4575486 c.1057G>A p.E353K Chr.12: 51292049-51292049 on Build GRCh38
694292 c.1030G>T p.A344S Chr.12: 51292076-51292076 on Build GRCh38
4665724 c.1029C>G p.P343P Chr.12: 51292077-51292077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777927634 Chr.12: 51291929-51291929 on Build GRCh38
rs772176957 Chr.12: 51292052-51292052 on Build GRCh38
rs369691754 Chr.12: 51292058-51292058 on Build GRCh38
rs138741584 Chr.12: 51292118-51292118 on Build GRCh38
rs748279114 Chr.12: 51292151-51292151 on Build GRCh38
rs550831260 Chr.12: 51292084-51292084 on Build GRCh38
rs200496180 Chr.12: 51291932-51291932 on Build GRCh38
rs750975779 Chr.12: 51291959-51291959 on Build GRCh38
rs763661034 Chr.12: 51292080-51292080 on Build GRCh38
rs141002466 Chr.12: 51292055-51292055 on Build GRCh38
rs776910520 Chr.12: 51292069-51292069 on Build GRCh38
rs921684243 Chr.12: 51291989-51291989 on Build GRCh38
rs376067729 Chr.12: 51292003-51292003 on Build GRCh38
rs140686836 Chr.12: 51291935-51291935 on Build GRCh38
rs201888825 Chr.12: 51291927-51291927 on Build GRCh38
rs931262941 Chr.12: 51291963-51291963 on Build GRCh38
rs142976170 Chr.12: 51292131-51292131 on Build GRCh38
rs112923160 Chr.12: 51292082-51292082 on Build GRCh38
rs761805381 Chr.12: 51291938-51291938 on Build GRCh38
rs145178409 Chr.12: 51291930-51291930 on Build GRCh38
rs1028688747 Chr.12: 51292072-51292072 on Build GRCh38
rs754220615 Chr.12: 51291984-51291984 on Build GRCh38
rs763228949 Chr.12: 51291934-51291934 on Build GRCh38
rs1056511629 Chr.12: 51292079-51292079 on Build GRCh38
rs895685933 Chr.12: 51292025-51292025 on Build GRCh38
rs771099064 Chr.12: 51291967-51291967 on Build GRCh38
rs777833539 Chr.12: 51291999-51291999 on Build GRCh38
rs61749591 Chr.12: 51291993-51291993 on Build GRCh38
rs746987882 Chr.12: 51292004-51292004 on Build GRCh38
rs376353509 Chr.12: 51292077-51292077 on Build GRCh38
rs188987098 Chr.12: 51292078-51292078 on Build GRCh38
rs897361854 Chr.12: 51292075-51292075 on Build GRCh38
rs201696311 Chr.12: 51291998-51291998 on Build GRCh38
rs759882503 Chr.12: 51292070-51292070 on Build GRCh38
rs965504112 Chr.12: 51292119-51292119 on Build GRCh38
rs374417427 Chr.12: 51292127-51292127 on Build GRCh38
rs766689090 Chr.12: 51291965-51291965 on Build GRCh38
rs757294613 Chr.12: 51292007-51292007 on Build GRCh38
rs745663605 Chr.12: 51292044-51292044 on Build GRCh38
rs781086442 Chr.12: 51292042-51292042 on Build GRCh38
rs910111500 Chr.12: 51292002-51292002 on Build GRCh38
rs146877980 Chr.12: 51291931-51291931 on Build GRCh38
rs748235442 Chr.12: 51292049-51292049 on Build GRCh38
rs766881298 Chr.12: 51292066-51292066 on Build GRCh38
rs965763447 Chr.12: 51291970-51291970 on Build GRCh38
rs757204229 Chr.12: 51292094-51292094 on Build GRCh38
rs774433785 Chr.12: 51291937-51291937 on Build GRCh38
rs760996900 Chr.12: 51291961-51291961 on Build GRCh38
rs150210505 Chr.12: 51292076-51292076 on Build GRCh38
rs774544875 Chr.12: 51292147-51292149 on Build GRCh38
rs975836597 Chr.12: 51291988-51291988 on Build GRCh38
rs752244584 Chr.12: 51291994-51291994 on Build GRCh38
rs375136926 Chr.12: 51291992-51291992 on Build GRCh38
rs1001650363 Chr.12: 51292000-51292000 on Build GRCh38
rs147701707 Chr.12: 51292097-51292097 on Build GRCh38
rs768859102 Chr.12: 51292045-51292045 on Build GRCh38
rs768053983 Chr.12: 51291945-51291945 on Build GRCh38
rs147443200 Chr.12: 51292149-51292149 on Build GRCh38
rs3210837 Chr.12: 51292047-51292047 on Build GRCh38

View Full Product Details