Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127712945-127713194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP157
Entrez Gene ID
286207
Gene Name
cilia and flagella associated protein 157
Gene Aliases
C9orf117
UniGene
Hs.652519
Gene Chromosome Location
Chr.9: 127706261-127716002 on Build GRCh38
Transcripts
NM_001012502.2 XM_006717064.3 XM_011518559.2 XM_017014630.1 XM_017014277.1

Gene Symbol
PTRH1
Entrez Gene ID
138428
Gene Name
peptidyl-tRNA hydrolase 1 homolog
Gene Aliases
C9orf115, PTH1
UniGene
Hs.653265
Gene Chromosome Location
Chr.9: 127695024-127716714 on Build GRCh38
Transcripts
NM_001012502.2 XM_006717064.3 XM_011518559.2 XM_017014630.1 XM_017014277.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6226728 c.1379G>T p.G460V Chr.9: 127713094-127713094 on Build GRCh38
5983376 c.1375C>T p.P459S Chr.9: 127713090-127713090 on Build GRCh38
7002568 c.1438C>A p.L480M Chr.9: 127713153-127713153 on Build GRCh38
7085773 c.1444T>C p.S482P Chr.9: 127713159-127713159 on Build GRCh38
1624571 c.1376_1377insG p.D461fs*37 Chr.9: 127713091-127713092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78968934 Chr.9: 127713092-127713092 on Build GRCh38
rs780458865 Chr.9: 127713108-127713108 on Build GRCh38
rs917241259 Chr.9: 127713157-127713157 on Build GRCh38
rs756490318 Chr.9: 127713023-127713023 on Build GRCh38
rs972437141 Chr.9: 127713013-127713013 on Build GRCh38
rs759050313 Chr.9: 127713082-127713082 on Build GRCh38
rs367607321 Chr.9: 127713118-127713118 on Build GRCh38
rs543912514 Chr.9: 127713051-127713051 on Build GRCh38
rs775800130 Chr.9: 127713017-127713017 on Build GRCh38
rs748671791 Chr.9: 127713122-127713122 on Build GRCh38
rs777719317 Chr.9: 127712986-127712986 on Build GRCh38
rs1025767068 Chr.9: 127713040-127713040 on Build GRCh38
rs761600893 Chr.9: 127713178-127713178 on Build GRCh38
rs575702552 Chr.9: 127713171-127713171 on Build GRCh38
rs935896423 Chr.9: 127712997-127712997 on Build GRCh38
rs960997152 Chr.9: 127713006-127713006 on Build GRCh38
rs375153055 Chr.9: 127712985-127712985 on Build GRCh38
rs541652643 Chr.9: 127712972-127712972 on Build GRCh38
rs374561061 Chr.9: 127713106-127713106 on Build GRCh38
rs755443452 Chr.9: 127713114-127713114 on Build GRCh38
rs746139886 Chr.9: 127713022-127713022 on Build GRCh38
rs752500714 Chr.9: 127712982-127712982 on Build GRCh38
rs780510350 Chr.9: 127713029-127713029 on Build GRCh38
rs750531847 Chr.9: 127713103-127713103 on Build GRCh38
rs763991791 Chr.9: 127713091-127713091 on Build GRCh38
rs200372951 Chr.9: 127713172-127713172 on Build GRCh38
rs757350739 Chr.9: 127713096-127713096 on Build GRCh38
rs781442675 Chr.9: 127713021-127713021 on Build GRCh38
rs34950677 Chr.9: 127713010-127713011 on Build GRCh38
rs776332587 Chr.9: 127713075-127713075 on Build GRCh38
rs746506232 Chr.9: 127713061-127713061 on Build GRCh38
rs901717083 Chr.9: 127713117-127713117 on Build GRCh38
rs751586501 Chr.9: 127713003-127713003 on Build GRCh38
rs766008254 Chr.9: 127712969-127712969 on Build GRCh38
rs371740113 Chr.9: 127713146-127713146 on Build GRCh38
rs572650181 Chr.9: 127712966-127712966 on Build GRCh38
rs79537388 Chr.9: 127713033-127713033 on Build GRCh38
rs759346417 Chr.9: 127712975-127712975 on Build GRCh38
rs374070528 Chr.9: 127713062-127713062 on Build GRCh38
rs769386392 Chr.9: 127713161-127713161 on Build GRCh38
rs770167060 Chr.9: 127712984-127712984 on Build GRCh38
rs370617973 Chr.9: 127712983-127712983 on Build GRCh38
rs199630039 Chr.9: 127713004-127713004 on Build GRCh38
rs764999071 Chr.9: 127712977-127712977 on Build GRCh38
rs497632 Chr.9: 127713163-127713163 on Build GRCh38
rs201886711 Chr.9: 127713112-127713112 on Build GRCh38
rs762756206 Chr.9: 127713085-127713085 on Build GRCh38
rs779533908 Chr.9: 127713115-127713115 on Build GRCh38
rs984895158 Chr.9: 127713072-127713072 on Build GRCh38
rs775021171 Chr.9: 127713053-127713053 on Build GRCh38
rs772645395 Chr.9: 127713138-127713138 on Build GRCh38
rs775224034 Chr.9: 127713083-127713083 on Build GRCh38
rs767555039 Chr.9: 127713101-127713101 on Build GRCh38

View Full Product Details