Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4698955-4699228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS3
Entrez Gene ID
124401
Gene Name
ankyrin repeat and sterile alpha motif domain containing 3
Gene Aliases
-
UniGene
Hs.654859
Gene Chromosome Location
Chr.16: 4696510-4734377 on Build GRCh38
Transcripts
NM_001242929.1 NM_001308089.1 NM_001324129.1 NM_001324130.1 NM_133450.3 NR_040252.1 XM_005255119.3 XM_005255121.3 XM_011522372.1 XM_011522373.1 XM_011522374.1 XM_011522378.1 XM_017022935.1 XM_017022936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5493382 c.1388A>G p.D463G Chr.16: 4699073-4699073 on Build GRCh38
4060848 c.1311C>T p.I437I Chr.16: 4699150-4699150 on Build GRCh38
2921187 c.1324T>C p.Y442H Chr.16: 4699137-4699137 on Build GRCh38
4579050 c.1409C>T p.T470M Chr.16: 4699052-4699052 on Build GRCh38
6322105 c.1311C>A p.I437I Chr.16: 4699150-4699150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774355243 Chr.16: 4699153-4699153 on Build GRCh38
rs778546392 Chr.16: 4699114-4699114 on Build GRCh38
rs185661020 Chr.16: 4699047-4699047 on Build GRCh38
rs770379990 Chr.16: 4698979-4698979 on Build GRCh38
rs775220732 Chr.16: 4699035-4699035 on Build GRCh38
rs371585380 Chr.16: 4699167-4699167 on Build GRCh38
rs751962073 Chr.16: 4699066-4699066 on Build GRCh38
rs759513353 Chr.16: 4698982-4698982 on Build GRCh38
rs922267266 Chr.16: 4698997-4698997 on Build GRCh38
rs768605878 Chr.16: 4699083-4699083 on Build GRCh38
rs769664244 Chr.16: 4699137-4699137 on Build GRCh38
rs369639876 Chr.16: 4699025-4699025 on Build GRCh38
rs568684634 Chr.16: 4699018-4699018 on Build GRCh38
rs145984825 Chr.16: 4699075-4699075 on Build GRCh38
rs760013486 Chr.16: 4699110-4699110 on Build GRCh38
rs924455911 Chr.16: 4698988-4698988 on Build GRCh38
rs181535369 Chr.16: 4699081-4699081 on Build GRCh38
rs767336753 Chr.16: 4698983-4698983 on Build GRCh38
rs750587696 Chr.16: 4699051-4699051 on Build GRCh38
rs781451809 Chr.16: 4699074-4699074 on Build GRCh38
rs148410195 Chr.16: 4699126-4699126 on Build GRCh38
rs1022994640 Chr.16: 4699080-4699080 on Build GRCh38
rs776004941 Chr.16: 4699207-4699207 on Build GRCh38
rs199645899 Chr.16: 4699203-4699203 on Build GRCh38
rs376181627 Chr.16: 4699036-4699036 on Build GRCh38
rs377633164 Chr.16: 4699111-4699111 on Build GRCh38
rs555313941 Chr.16: 4699190-4699190 on Build GRCh38
rs537329747 Chr.16: 4699040-4699040 on Build GRCh38
rs138763631 Chr.16: 4699100-4699100 on Build GRCh38
rs772675026 Chr.16: 4698975-4698975 on Build GRCh38
rs754052823 Chr.16: 4698993-4698993 on Build GRCh38
rs142579917 Chr.16: 4699072-4699072 on Build GRCh38
rs745373264 Chr.16: 4699029-4699029 on Build GRCh38
rs747405935 Chr.16: 4699019-4699019 on Build GRCh38
rs375152022 Chr.16: 4699023-4699023 on Build GRCh38
rs114131563 Chr.16: 4699208-4699208 on Build GRCh38
rs778536183 Chr.16: 4699076-4699076 on Build GRCh38
rs776615282 Chr.16: 4699096-4699096 on Build GRCh38
rs887284400 Chr.16: 4699069-4699069 on Build GRCh38
rs755240576 Chr.16: 4699021-4699021 on Build GRCh38
rs750497675 Chr.16: 4699010-4699010 on Build GRCh38
rs373370187 Chr.16: 4699026-4699026 on Build GRCh38
rs528487941 Chr.16: 4698994-4698994 on Build GRCh38
rs373937147 Chr.16: 4699101-4699101 on Build GRCh38
rs772118388 Chr.16: 4699033-4699033 on Build GRCh38
rs749098997 Chr.16: 4699200-4699200 on Build GRCh38
rs766039619 Chr.16: 4699191-4699191 on Build GRCh38
rs201848967 Chr.16: 4698987-4698987 on Build GRCh38
rs987015760 Chr.16: 4699194-4699194 on Build GRCh38
rs377144011 Chr.16: 4699002-4699002 on Build GRCh38
rs748900379 Chr.16: 4699024-4699024 on Build GRCh38
rs144602666 Chr.16: 4699123-4699123 on Build GRCh38
rs766923282 Chr.16: 4699062-4699062 on Build GRCh38
rs762165596 Chr.16: 4699041-4699041 on Build GRCh38
rs200458576 Chr.16: 4699168-4699168 on Build GRCh38
rs773600265 Chr.16: 4699141-4699141 on Build GRCh38
rs141891119 Chr.16: 4699105-4699105 on Build GRCh38
rs761046094 Chr.16: 4699174-4699174 on Build GRCh38
rs754196652 Chr.16: 4699188-4699188 on Build GRCh38
rs777479053 Chr.16: 4699199-4699199 on Build GRCh38
rs955609941 Chr.16: 4698999-4698999 on Build GRCh38
rs747960894 Chr.16: 4699135-4699135 on Build GRCh38
rs370773868 Chr.16: 4699017-4699017 on Build GRCh38
rs375010906 Chr.16: 4699197-4699197 on Build GRCh38
rs751096575 Chr.16: 4699192-4699192 on Build GRCh38
rs557129144 Chr.16: 4698986-4698986 on Build GRCh38
rs940165376 Chr.16: 4699071-4699071 on Build GRCh38
rs914369052 Chr.16: 4699008-4699008 on Build GRCh38
rs771421144 Chr.16: 4699078-4699078 on Build GRCh38
rs201052203 Chr.16: 4699006-4699006 on Build GRCh38
rs113252244 Chr.16: 4699107-4699107 on Build GRCh38
rs762936052 Chr.16: 4699061-4699061 on Build GRCh38
rs200504243 Chr.16: 4698976-4698976 on Build GRCh38
rs552702968 Chr.16: 4698981-4698981 on Build GRCh38
rs776917162 Chr.16: 4699211-4699211 on Build GRCh38
rs746877380 Chr.16: 4699082-4699082 on Build GRCh38
rs772086923 Chr.16: 4699206-4699206 on Build GRCh38
rs758034712 Chr.16: 4699189-4699189 on Build GRCh38
rs766331417 Chr.16: 4699102-4699102 on Build GRCh38
rs746504268 Chr.16: 4699129-4699129 on Build GRCh38
rs771163966 Chr.16: 4699150-4699150 on Build GRCh38
rs764560113 Chr.16: 4699210-4699210 on Build GRCh38
rs745383444 Chr.16: 4699077-4699077 on Build GRCh38
rs974876814 Chr.16: 4699156-4699156 on Build GRCh38
rs368102357 Chr.16: 4699201-4699201 on Build GRCh38
rs764399022 Chr.16: 4699183-4699183 on Build GRCh38
rs768532604 Chr.16: 4699130-4699130 on Build GRCh38
rs756089105 Chr.16: 4699198-4699198 on Build GRCh38
rs745951407 Chr.16: 4699204-4699204 on Build GRCh38
rs775915015 Chr.16: 4699169-4699169 on Build GRCh38
rs200161297 Chr.16: 4699134-4699134 on Build GRCh38

View Full Product Details