Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 121820910-121821171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNA2
Entrez Gene ID
890
Gene Name
cyclin A2
Gene Aliases
CCN1, CCNA
UniGene
Hs.58974
Gene Chromosome Location
Chr.4: 121816444-121823933 on Build GRCh38
Transcripts
NM_001237.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1618341 c.458-10A>T p.? Chr.4: 121821101-121821101 on Build GRCh38
3825084 c.567G>T p.M189I Chr.4: 121820982-121820982 on Build GRCh38
1247602 c.549C>A p.H183Q Chr.4: 121821000-121821000 on Build GRCh38
5797950 c.529G>A p.D177N Chr.4: 121821020-121821020 on Build GRCh38
5425608 c.487A>G p.I163V Chr.4: 121821062-121821062 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751264133 Chr.4: 121820951-121820951 on Build GRCh38
rs916333563 Chr.4: 121821008-121821008 on Build GRCh38
rs758147266 Chr.4: 121821121-121821122 on Build GRCh38
rs201310057 Chr.4: 121820972-121820972 on Build GRCh38
rs981753726 Chr.4: 121821013-121821013 on Build GRCh38
rs368094973 Chr.4: 121821041-121821041 on Build GRCh38
rs777870825 Chr.4: 121821014-121821014 on Build GRCh38
rs909697729 Chr.4: 121821095-121821095 on Build GRCh38
rs756376701 Chr.4: 121821126-121821126 on Build GRCh38
rs754673374 Chr.4: 121821114-121821114 on Build GRCh38
rs764812450 Chr.4: 121821076-121821076 on Build GRCh38
rs752882999 Chr.4: 121821124-121821124 on Build GRCh38
rs772496110 Chr.4: 121821065-121821065 on Build GRCh38
rs761627971 Chr.4: 121821071-121821071 on Build GRCh38
rs578250642 Chr.4: 121821147-121821147 on Build GRCh38
rs370029186 Chr.4: 121821139-121821139 on Build GRCh38
rs375777201 Chr.4: 121821134-121821134 on Build GRCh38
rs757358346 Chr.4: 121821140-121821140 on Build GRCh38
rs913425988 Chr.4: 121821080-121821080 on Build GRCh38
rs147878070 Chr.4: 121821010-121821010 on Build GRCh38
rs750385370 Chr.4: 121820942-121820942 on Build GRCh38
rs774950274 Chr.4: 121821044-121821044 on Build GRCh38
rs769241 Chr.4: 121821142-121821142 on Build GRCh38
rs750203991 Chr.4: 121821101-121821102 on Build GRCh38
rs751172069 Chr.4: 121821135-121821139 on Build GRCh38
rs776412937 Chr.4: 121821070-121821070 on Build GRCh38
rs766175146 Chr.4: 121820945-121820945 on Build GRCh38
rs769242 Chr.4: 121821062-121821062 on Build GRCh38
rs771290221 Chr.4: 121821042-121821042 on Build GRCh38
rs777992077 Chr.4: 121821127-121821127 on Build GRCh38
rs754835875 Chr.4: 121820958-121820958 on Build GRCh38
rs762548324 Chr.4: 121821096-121821096 on Build GRCh38
rs142481279 Chr.4: 121821037-121821037 on Build GRCh38
rs766013419 Chr.4: 121821124-121821127 on Build GRCh38
rs772900561 Chr.4: 121821083-121821083 on Build GRCh38
rs767365609 Chr.4: 121821120-121821120 on Build GRCh38
rs758310981 Chr.4: 121820943-121820943 on Build GRCh38
rs781326696 Chr.4: 121820964-121820964 on Build GRCh38
rs5031014 Chr.4: 121821103-121821103 on Build GRCh38
rs938962336 Chr.4: 121821040-121821040 on Build GRCh38
rs751521188 Chr.4: 121821112-121821112 on Build GRCh38

View Full Product Details