Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27578553-27578814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf16
Entrez Gene ID
84226
Gene Name
chromosome 2 open reading frame 16
Gene Aliases
-
UniGene
Hs.131021
Gene Chromosome Location
Chr.2: 27576522-27582722 on Build GRCh38
Transcripts
NM_032266.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
419208 c.2003C>T p.S668F Chr.2: 27578575-27578575 on Build GRCh38
3910162 c.2174G>A p.G725E Chr.2: 27578746-27578746 on Build GRCh38
1668811 c.2234G>T p.R745I Chr.2: 27578806-27578806 on Build GRCh38
2940368 c.2029A>G p.T677A Chr.2: 27578601-27578601 on Build GRCh38
6158092 c.2213C>T p.T738I Chr.2: 27578785-27578785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754495342 Chr.2: 27578593-27578593 on Build GRCh38
rs1919127 Chr.2: 27578626-27578626 on Build GRCh38
rs777549367 Chr.2: 27578702-27578702 on Build GRCh38
rs929944623 Chr.2: 27578761-27578761 on Build GRCh38
rs896421360 Chr.2: 27578732-27578732 on Build GRCh38
rs182932260 Chr.2: 27578779-27578779 on Build GRCh38
rs780573516 Chr.2: 27578705-27578705 on Build GRCh38
rs767412185 Chr.2: 27578648-27578649 on Build GRCh38
rs373833265 Chr.2: 27578704-27578704 on Build GRCh38
rs773407710 Chr.2: 27578747-27578747 on Build GRCh38
rs747493329 Chr.2: 27578722-27578722 on Build GRCh38
rs747766823 Chr.2: 27578597-27578597 on Build GRCh38
rs374774445 Chr.2: 27578709-27578709 on Build GRCh38
rs769964157 Chr.2: 27578745-27578745 on Build GRCh38
rs751492134 Chr.2: 27578657-27578657 on Build GRCh38
rs752671792 Chr.2: 27578653-27578670 on Build GRCh38
rs776589579 Chr.2: 27578616-27578616 on Build GRCh38
rs766218324 Chr.2: 27578639-27578639 on Build GRCh38
rs748146608 Chr.2: 27578735-27578735 on Build GRCh38
rs755748902 Chr.2: 27578695-27578695 on Build GRCh38
rs369834274 Chr.2: 27578693-27578693 on Build GRCh38
rs368826862 Chr.2: 27578787-27578787 on Build GRCh38
rs752166511 Chr.2: 27578694-27578694 on Build GRCh38
rs761334012 Chr.2: 27578613-27578613 on Build GRCh38
rs774559737 Chr.2: 27578714-27578714 on Build GRCh38
rs749011418 Chr.2: 27578703-27578703 on Build GRCh38
rs779934792 Chr.2: 27578590-27578590 on Build GRCh38
rs780633070 Chr.2: 27578595-27578595 on Build GRCh38
rs769066275 Chr.2: 27578728-27578728 on Build GRCh38
rs762046187 Chr.2: 27578617-27578617 on Build GRCh38
rs750682286 Chr.2: 27578636-27578636 on Build GRCh38
rs772618198 Chr.2: 27578611-27578611 on Build GRCh38
rs763228385 Chr.2: 27578756-27578756 on Build GRCh38
rs77286755 Chr.2: 27578658-27578658 on Build GRCh38
rs779367550 Chr.2: 27578600-27578600 on Build GRCh38
rs539117048 Chr.2: 27578610-27578610 on Build GRCh38
rs751547135 Chr.2: 27578591-27578591 on Build GRCh38
rs768716266 Chr.2: 27578615-27578615 on Build GRCh38
rs776734014 Chr.2: 27578730-27578730 on Build GRCh38

View Full Product Details