Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103520486-103520733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKB
Entrez Gene ID
1152
Gene Name
creatine kinase B
Gene Aliases
B-CK, BCK, CKBB, HEL-211, HEL-S-29
UniGene
Hs.173724
Gene Chromosome Location
Chr.14: 103519658-103522859 on Build GRCh38
Transcripts
NM_001823.4 XM_017020951.1 XM_017020952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1128327 c.661G>T p.D221Y Chr.14: 103520585-103520585 on Build GRCh38
2025552 c.731delG p.G244fs*3 Chr.14: 103520515-103520515 on Build GRCh38
5965680 c.723G>A p.Q241Q Chr.14: 103520523-103520523 on Build GRCh38
6298129 c.671C>A p.T224N Chr.14: 103520575-103520575 on Build GRCh38
4167720 c.755G>T p.R252L Chr.14: 103520491-103520491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201488531 Chr.14: 103520579-103520579 on Build GRCh38
rs530669346 Chr.14: 103520631-103520631 on Build GRCh38
rs767405482 Chr.14: 103520517-103520517 on Build GRCh38
rs780589580 Chr.14: 103520571-103520571 on Build GRCh38
rs760568631 Chr.14: 103520615-103520615 on Build GRCh38
rs757292875 Chr.14: 103520640-103520640 on Build GRCh38
rs766252400 Chr.14: 103520619-103520619 on Build GRCh38
rs528121888 Chr.14: 103520543-103520543 on Build GRCh38
rs756513630 Chr.14: 103520644-103520644 on Build GRCh38
rs373038236 Chr.14: 103520520-103520520 on Build GRCh38
rs764289385 Chr.14: 103520634-103520634 on Build GRCh38
rs749765560 Chr.14: 103520574-103520574 on Build GRCh38
rs1054583106 Chr.14: 103520687-103520687 on Build GRCh38
rs550759782 Chr.14: 103520639-103520639 on Build GRCh38
rs758597982 Chr.14: 103520537-103520537 on Build GRCh38
rs887679511 Chr.14: 103520561-103520561 on Build GRCh38
rs199734997 Chr.14: 103520556-103520556 on Build GRCh38
rs772882430 Chr.14: 103520614-103520614 on Build GRCh38
rs746220920 Chr.14: 103520568-103520568 on Build GRCh38
rs998845951 Chr.14: 103520678-103520678 on Build GRCh38
rs771627562 Chr.14: 103520611-103520611 on Build GRCh38
rs141472120 Chr.14: 103520583-103520583 on Build GRCh38
rs148243589 Chr.14: 103520505-103520505 on Build GRCh38
rs200697777 Chr.14: 103520641-103520641 on Build GRCh38
rs773723702 Chr.14: 103520596-103520596 on Build GRCh38
rs747636497 Chr.14: 103520597-103520597 on Build GRCh38
rs757404572 Chr.14: 103520544-103520544 on Build GRCh38
rs759542035 Chr.14: 103520632-103520632 on Build GRCh38
rs934282235 Chr.14: 103520637-103520637 on Build GRCh38
rs559489078 Chr.14: 103520519-103520519 on Build GRCh38

View Full Product Details