Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66800548-66800787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf80
Entrez Gene ID
79703
Gene Name
chromosome 11 open reading frame 80
Gene Aliases
TOP6BL, TOPOVIBL
UniGene
Hs.292088
Gene Chromosome Location
Chr.11: 66744736-66843516 on Build GRCh38
Transcripts
NM_001302084.1 NM_024650.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7193865 c.746A>G p.N249S Chr.11: 66800669-66800669 on Build GRCh38
7195293 c.766A>G p.I256V Chr.11: 66800689-66800689 on Build GRCh38
6333822 c.718-4G>T p.? Chr.11: 66800637-66800637 on Build GRCh38
3869966 c.727C>T p.H243Y Chr.11: 66800650-66800650 on Build GRCh38
7400307 c.732C>T p.F244F Chr.11: 66800655-66800655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768108649 Chr.11: 66800602-66800602 on Build GRCh38
rs369191326 Chr.11: 66800582-66800582 on Build GRCh38
rs542244777 Chr.11: 66800636-66800636 on Build GRCh38
rs755187009 Chr.11: 66800645-66800645 on Build GRCh38
rs1026003343 Chr.11: 66800700-66800700 on Build GRCh38
rs781019380 Chr.11: 66800683-66800683 on Build GRCh38
rs750903107 Chr.11: 66800617-66800617 on Build GRCh38
rs779020740 Chr.11: 66800646-66800646 on Build GRCh38
rs745766170 Chr.11: 66800689-66800689 on Build GRCh38
rs191651158 Chr.11: 66800676-66800676 on Build GRCh38
rs761442553 Chr.11: 66800728-66800728 on Build GRCh38
rs373448823 Chr.11: 66800630-66800630 on Build GRCh38
rs751103343 Chr.11: 66800596-66800596 on Build GRCh38
rs1008061430 Chr.11: 66800638-66800638 on Build GRCh38
rs769485214 Chr.11: 66800695-66800695 on Build GRCh38
rs774045692 Chr.11: 66800719-66800719 on Build GRCh38
rs763766984 Chr.11: 66800592-66800592 on Build GRCh38
rs762755722 Chr.11: 66800713-66800713 on Build GRCh38
rs768404905 Chr.11: 66800714-66800714 on Build GRCh38
rs754011225 Chr.11: 66800632-66800632 on Build GRCh38
rs756458009 Chr.11: 66800622-66800622 on Build GRCh38
rs770893070 Chr.11: 66800680-66800680 on Build GRCh38
rs890105934 Chr.11: 66800701-66800701 on Build GRCh38
rs766295177 Chr.11: 66800620-66800624 on Build GRCh38
rs762460176 Chr.11: 66800599-66800599 on Build GRCh38
rs376103610 Chr.11: 66800709-66800709 on Build GRCh38

View Full Product Details