Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50872248-50872479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD12B
Entrez Gene ID
145447
Gene Name
abhydrolase domain containing 12B
Gene Aliases
BEM46L3, C14orf29, c14_5314
UniGene
Hs.271896
Gene Chromosome Location
Chr.14: 50872090-50904970 on Build GRCh38
Transcripts
NM_001206673.1 NM_181533.3 NM_181814.1 XM_006720052.1 XM_011536472.1 XM_011536474.2 XM_017021031.1

SNP Detail

SNP ID Chromosome Location
rs894982509 Chr.14: 50872386-50872386 on Build GRCh38
rs1054396955 Chr.14: 50872310-50872310 on Build GRCh38
rs181194043 Chr.14: 50872346-50872346 on Build GRCh38
rs745909978 Chr.14: 50872279-50872279 on Build GRCh38
rs187100872 Chr.14: 50872360-50872360 on Build GRCh38
rs889492213 Chr.14: 50872307-50872307 on Build GRCh38
rs922288021 Chr.14: 50872283-50872283 on Build GRCh38
rs998889912 Chr.14: 50872419-50872419 on Build GRCh38
rs1028912029 Chr.14: 50872309-50872309 on Build GRCh38
rs12894798 Chr.14: 50872336-50872336 on Build GRCh38
rs1049498746 Chr.14: 50872299-50872299 on Build GRCh38
rs954176996 Chr.14: 50872320-50872320 on Build GRCh38
rs906541157 Chr.14: 50872342-50872342 on Build GRCh38
rs1043186277 Chr.14: 50872396-50872396 on Build GRCh38
rs904593392 Chr.14: 50872402-50872402 on Build GRCh38
rs150967377 Chr.14: 50872410-50872411 on Build GRCh38
rs779097718 Chr.14: 50872278-50872278 on Build GRCh38
rs772753106 Chr.14: 50872291-50872291 on Build GRCh38
rs190038452 Chr.14: 50872379-50872379 on Build GRCh38
rs933627545 Chr.14: 50872299-50872300 on Build GRCh38
rs966005055 Chr.14: 50872296-50872296 on Build GRCh38
rs757049135 Chr.14: 50872331-50872331 on Build GRCh38
rs972635871 Chr.14: 50872414-50872414 on Build GRCh38
rs541751997 Chr.14: 50872394-50872394 on Build GRCh38
rs747398383 Chr.14: 50872304-50872304 on Build GRCh38
rs909617638 Chr.14: 50872338-50872338 on Build GRCh38
rs768994408 Chr.14: 50872312-50872312 on Build GRCh38
rs922487583 Chr.14: 50872438-50872438 on Build GRCh38
rs553556161 Chr.14: 50872368-50872368 on Build GRCh38
rs904315582 Chr.14: 50872420-50872420 on Build GRCh38
rs776050870 Chr.14: 50872303-50872303 on Build GRCh38
rs961174195 Chr.14: 50872404-50872404 on Build GRCh38
rs190024469 Chr.14: 50872273-50872273 on Build GRCh38
rs933658782 Chr.14: 50872459-50872459 on Build GRCh38
rs12894955 Chr.14: 50872407-50872407 on Build GRCh38
rs557043824 Chr.14: 50872329-50872329 on Build GRCh38
rs533733022 Chr.14: 50872384-50872384 on Build GRCh38

View Full Product Details